missing translation for 'onlineSavingsMsg'
Learn More

CD278 (ICOS), PE-Cyanine7, clone: 7E.17G9, eBioscience™

Rat Monoclonal Antibody

Brand:  Affymetrix eBioscience 25-9942-80

Code : NEW

Additional Details : Weight : 0.09500kg

 View more versions of this product

Product Code. 15528696

  • £70.03 / 25µg


EU Stock 2
Add to basket

Description

Description

Description: The 7E.17G9 monoclonal antibody reacts with mouse ICOS (Inducible COStimulatory molecule), a T cell specific molecule and a third member of the CD28/CTLA-4 family. A homodimer of 47-57 kDa, ICOS is expressed on activated T cells, has potent costimulatory activity for T cell activation and proliferation and is required for humoral immune response. ICOS binds to its ligand on activated APC including B cells called B7h/B7RP-1 and is thought to play a protective role in inflammatory autoimmune diseases. ICOS may be involved in the development of Th2 cells.Applications Reported: This 7E.17G9 antibody has been reported for use in flow cytometric analysis.Applications Tested: This 7E.17G9 antibody has been tested by flow cytometric analysis of stimulated mouse splenocytes. This can be used at less than or equal to 0.25 μg per test. A test is defined as the amount (μg) of antibody that will stain a cell sample in a final volume of 100 μL. Cell number should be determined empirically but can range from 10^5 to 10^8 cells/test. It is recommended that the antibody be carefully titrated for optimal performance in the assay of interest.Light sensitivity: This tandem dye is sensitive photo-induced oxidation. Please protect this vial and stained samples from light. Fixation: Samples can be stored in IC Fixation Buffer (cat. 00-8222) (100 μL cell sample + 100 μL IC Fixation Buffer) or 1-step Fix/Lyse Solution (cat. 00-5333) for up to 3 days in the dark at 4°C with minimal impact on brightness and FRET efficiency/compensation. Some generalizations regarding fluorophore performance after fixation can be made, but clone specific performance should be determined empirically.Excitation: 488-561 nm; Emission: 775 nm; Laser: Blue Laser, Green Laser, Yellow-Green Laser.Filtration: 0.2μm post-manufacturing filtered.
Specifications

Specifications

CD278 (ICOS)
Monoclonal
0.2mg/mL
PBS with 0.1% gelatin and 0.09% sodium azide; pH 7.2
Q9WVS0
AILIM, CD278, CVID1, activation-inducible lymphocyte immunomediatory molecule, inducible T-cell costimulator, inducible costimulator
Rat
Affinity chromatography
RUO
Antibody
Monoclonal
Murine
Flow Cytometry
7E.17G9
PE-Cyanine7
Icos
Conjugated
Icos
IgG2b, kappa
25μg
Store at 2-8°C. Do not freeze. Light-sensitive material. This tandem dye is sensitive to photo-induced oxidation. Protect this vial from light during storage, handling and experimental procedures.
Primary
54167
Videos

For Research Use Only.