missing translation for 'onlineSavingsMsg'
Learn More

CD278 (ICOS), Functional Grade, clone: 7E.17G9, eBioscience™

Rat Monoclonal Antibody

Brand:  Affymetrix eBioscience 16-9942-81

Code : Z2

Additional Details : Weight : 0.01000kg

 View more versions of this product

Product Code. 15208847

  • £68.45 / 50µg


Call for latest availability
+44 (0)1509 555 500
Add to basket

Description

Description

Description: The 7E.17G9 monoclonal antibody reacts with mouse ICOS (Inducible COStimulatory molecule), a T cell specific molecule and a third member of the CD28/CTLA-4 family. A homodimer of 47-57 kDa, ICOS is expressed on activated T cells, has potent costimulatory activity for T cell activation and proliferation and is required for humoral immune response. ICOS binds to its ligand on activated APC including B cells called B7h/B7RP-1 and is thought to play a protective role in inflammatory autoimmune diseases. ICOS may be involved in the development of Th2 cells.Applications Reported: 7E.17G9 has been reported for use in flow cytometric analysis and for inhibition of ligand binding in functional assays.Applications Tested: The 7E.17G9 antibody has been tested by flow cytometric analysis of unstimulated and Con A activated (3 day) mouse splenocytes and can be used at less than or equal to 0.125 μg per test. A test is defined as the amount (μg) of antibody that will stain a cell sample in a final volume of 100 μL. Cell number should be determined empirically but can range from 10^5 to 10^8 cells/test. It is recommended that the antibody be carefully titrated for optimal performance in the assay of interest.Endotoxin: Less than 0.001 ng/ug antibody as determined by the LAL assay.Storage and handling: Use in a sterile environment.Filtration: 0.2μm post-manufacturing filtered.Purity: Greater than 90%, as determined by SDS-PAGE.Endotoxin Level: Less than 0.001 ng/μg antibody, as determined by LAL assay.Aggregation: Less than 10%, as determined by HPLC.
Specifications

Specifications

CD278 (ICOS)
Monoclonal
1mg/mL
PBS with no preservative; pH 7.2
Q9WVS0
AILIM, CD278, CVID1, activation-inducible lymphocyte immunomediatory molecule, inducible T-cell costimulator, inducible costimulator
Rat
Affinity chromatography
RUO
Antibody
Monoclonal
Murine
Functional Assay
7E.17G9
Functional Grade
Icos
Purified
Icos
IgG2b, kappa
50μg
Store at 2-8°C.
Primary
54167
Videos

For Research Use Only.