missing translation for 'onlineSavingsMsg'
Learn More

CD276 (B7-H3), APC, clone: 7-517, eBioscience™

Mouse Monoclonal Antibody

Brand:  Affymetrix eBioscience 17-2769-42

Code : NEW

Additional Details : Weight : 0.23750kg

 View more versions of this product

Product Code. 15588066

  • £174.50 / 100 tests


Estimated Shipment date
from Supplier 22-01-2018
Add to basket

Disclaimers:

For Research Use Only.


Description

Description

Description: This 7-517 monoclonal antibody reacts with human CD276, which is also known as B7-H3. This type I transmembrane protein is expressed on immature and mature dendritic cells but not monocytes, granulocytes, or resting lymphocytes. Moreover, CD276 is expressed on non-hematopoietic cells such as osteoblasts, fibroblasts, and epithelial cells. However, CD276 expression can be induced on dendritic cells, monocytes, T, B, and NK cells. This co-stimulatory molecule binds activated T cells, leading to cell proliferation and IFN-gamma production.Applications Reported: This 7-517 antibody has been reported for use in flow cytometric analysis.Applications Tested: This 7-517 antibody has been pre-titrated and tested by flow cytometric analysis of normal human monocyte-derived dendritic cells. This can be used at 5 μL (0.125 μg) per test. A test is defined as the amount (μg) of antibody that will stain a cell sample in a final volume of 100 μL. Cell number should be determined empirically but can range from 10^5 to 10^8 cells/test.Excitation: 633-647 nm; Emission: 660 nm; Laser: Red Laser.Filtration: 0.2μm post-manufacturing filtered.
Specifications

Specifications

CD276
7-517
APC
Conjugated
Murine
100 Tests
Store at 2-8°C. Do not freeze. Light-sensitive material.
Monoclonal
Flow Cytometry
5μL (0.125μg)/test
aqueous buffer, 0.09% sodium azide, may contain carrier protein/stabilizer
B7 homolog 3
IgG1
RUO
Primary
Human
Videos