missing translation for 'onlineSavingsMsg'
Learn More

CD276 (B7-H3), APC, clone: 7-517, eBioscience™

Mouse Monoclonal Antibody

Brand:  Affymetrix eBioscience 17-2769-41

Code : NEW

Additional Details : Weight : 0.23750kg

Product Code. 15578066

  • £72.00 / 25 tests

Estimated Shipment
04-11-2020

Please to purchase this item. Need a web account? Register with us today!


Description

Description

Description: This 7-517 monoclonal antibody reacts with human CD276, which is also known as B7-H3. This type I transmembrane protein is expressed on immature and mature dendritic cells but not monocytes, granulocytes, or resting lymphocytes. Moreover, CD276 is expressed on non-hematopoietic cells such as osteoblasts, fibroblasts, and epithelial cells. However, CD276 expression can be induced on dendritic cells, monocytes, T, B, and NK cells. This co-stimulatory molecule binds activated T cells, leading to cell proliferation and IFN-gamma production.Applications Reported: This 7-517 antibody has been reported for use in flow cytometric analysis.Applications Tested: This 7-517 antibody has been pre-titrated and tested by flow cytometric analysis of normal human monocyte-derived dendritic cells. This can be used at 5 μL (0.125 μg) per test. A test is defined as the amount (μg) of antibody that will stain a cell sample in a final volume of 100 μL. Cell number should be determined empirically but can range from 10^5 to 10^8 cells/test.Excitation: 633-647 nm; Emission: 660 nm; Laser: Red Laser.Filtration: 0.2μm post-manufacturing filtered.
Specifications

Specifications

CD276 (B7-H3)
Monoclonal
5μl/Test
PBS with 0.1% gelatin, 0.2% BSA and 0.09% sodium azide; pH 7.2
Q5ZPR3
PSEC0249, 4Ig-B7-H3, B7-H3, B7H3, B7RP-2, B7 homolog 3, CD276 antigen, costimulatory molecule
Murine
Affinity chromatography
RUO
Antibody
Monoclonal
Human
Flow Cytometry
7-517
APC
CD276
Liquid
CD276
IgG1, kappa, kappa
25 Tests
Store at 2-8°C. Do not freeze. Light-sensitive material.
Primary
80381
Videos
Documents

For Research Use Only.