Learn More
CD276 (B7-H3), PerCP-eFluor™ 710, clone: 7-517, eBioscience™
Mouse Monoclonal Antibody
Brand:  Affymetrix eBioscience 46-2769-41
Code : NEW
Additional Details : Weight : 0.23750kg
 View more versions of this product
Product Code. 15579226
  • £66.65 / 25 tests
Estimated Shipment date
from Supplier 26-03-2020
Description
Description
This 7-517 monoclonal antibody reacts with human CD276, which is also known as B7-H3. This type I transmembrane protein is expressed on immature and mature dendritic cells but not monocytes, granulocytes, or resting lymphocytes. Moreover, CD276 is expressed on non-hematopoietic cells such as osteoblasts, fibroblasts, and epithelial cells. However, CD276 expression can be induced on dendritic cells, monocytes, T, B, and NK cells. This co-stimulatory molecule binds activated T cells, leading to cell proliferation and IFN-gamma production.
Specifications
Specifications
CD276 (B7-H3)
7-517
PerCP-eFluor 710
Conjugated
Murine
25 Tests
Store at 2-8°C. Do not freeze. Light-sensitive material. This tandem dye is sensitive to photo-induced oxidation. Protect this vial from light during storage, handling and experimental procedures.
Monoclonal
Flow Cytometry
5μL (0.125μg)/test
aqueous buffer, 0.09% sodium azide, may contain carrier protein/stabilizer
B7 homolog 3
IgG1, kappa