Learn More
CD275 (B7-H2) Monoclonal Antibody (MIH12), PE, eBioscience™, Invitrogen™
Mouse Monoclonal Antibody
£182.00
Specifications
Antigen | CD275 (B7-H2) |
---|---|
Clone | MIH12 |
Concentration | 5 μL/Test |
Applications | Flow Cytometry |
Classification | Monoclonal |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
15218031
|
Affymetrix eBioscience
12-5889-42 |
100 Tests |
£182.00
100 tests |
Estimated Shipment: 08-05-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
Description
Description: The MIH12 monoclonal antibody reacts with human B7RP-1, also known as B7h, B7-H2, GL50 and ICOS Ligand. B7RP-1, a member of the B7 family, has a predicted molecular weight of approximately 40 kDa and belongs to the Ig superfamily. Human B7RP-1 is expressed by activated monocytes/macrophages. B7RP-1 binds to the ICOS molecule (AILIM, CRP-1) expressed by activated T cells. The interaction of ICOS/B7RP-1 plays an important role in the T cell costimulation pathway. Applications Reported: The MIH12 antibody has been reported for use in flow cytometric analysis. Applications Tested: This MIH12 antibody has been pre-titrated and tested by flow cytometric analysis of normal human peripheral blood monocyte-derived dendritic cells. This can be used at 5 μL (0.125 μg) per test. A test is defined as the amount (μg) of antibody that will stain a cell sample in a final volume of 100 μL. Cell number should be determined empirically but can range from 10^5 to 10^8 cells/test. Excitation: 488-561 nm; Emission: 578 nm; Laser: Blue Laser, Green Laser, Yellow-Green Laser. Filtration: 0.2 μm post-manufacturing filtered.
Inducible co-stimulator ligand (ICOSL) is a specific ligand on antigen-presenting cells and cells of the peripheral tissue that binds to the inducible co-stimulator receptor (ICOS). ICOS belongs to the CD28/CD152 receptor family that regulates T-cell activation and function. ICOSL is expressed on monocytes, dendritic cells and B cells and can be induced by inflammatory stimuli in peripheral tissue. Binding to ICOSL delivers a co-stimulatory signal for T cell proliferation and cytokine secretion.Specifications
CD275 (B7-H2) | |
5 μL/Test | |
Monoclonal | |
Liquid | |
RUO | |
O75144 | |
ICOSLG | |
Primary | |
4° C, store in dark, DO NOT FREEZE! | |
ICOSLG |
MIH12 | |
Flow Cytometry | |
PE | |
Mouse | |
Human | |
23308 | |
IgG1 κ | |
Affinity chromatography | |
Antibody |
For Research Use Only.