missing translation for 'onlineSavingsMsg'
Learn More

CD275 (B7-H2), Biotin, clone: MIH12, eBioscience™

Mouse Monoclonal Antibody

Brand:  Affymetrix eBioscience 13-5889-80

Code : Z2

Additional Details : Weight : 0.25000kg

 View more versions of this product

Product Code. 13455289

  • £56.31 / 25Āµg


EU Stock 4
Add to basket

Description

Description

Description: The MIH12 monoclonal antibody reacts with human B7RP-1, also known as B7h, B7-H2, GL50 and ICOS Ligand. B7RP-1, a member of the B7 family, has a predicted molecular weight of approximately 40kDa and belongs to the Ig superfamily. Human B7RP-1 is expressed by activated monocytes/macrophages. B7RP-1 binds to the ICOS molecule (AILIM, CRP-1) expressed by activated T cells. The interaction of ICOS/B7RP-1 plays an important role in the T cell costimulation pathway.Applications Reported: This MIH12 antibody has been reported for use in flow cytometric analysis, and immunohistology staining of frozen tissue sections. Applicability of MIH12 for immunoprecipitation and immunoblotting (WB) has not been determined.Applications Tested: The MIH12 antibody has been tested by flow cytometric analysis of normal human peripheral blood monocyte-derived dendritic cells. This can be used at less than or equal to 1 μg per test. A test is defined as the amount (μg) of antibody that will stain a cell sample in a final volume of 100 μL. Cell number should be determined empirically but can range from 10^5 to 10^8 cells/test. It is recommended that the antibody be carefully titrated for optimal performance in the assay of interest.Filtration: 0.2μm post-manufacturing filtered.
Specifications

Specifications

CD275 (B7-H2)
Monoclonal
0.5mg/mL
PBS with 0.1% gelatin and 0.09% sodium azide; pH 7.2
O75144
B7-H2, B7H2, B7RP-1, B7RP1, CD275, GL50, ICOS-L, ICOSL, KIAA0653, LICOS, ICOS Ligand
Murine
Affinity chromatography
RUO
Antibody
Monoclonal
Human
Flow CytometryĆ¾Immunohistochemistry (Frozen)
MIH12
Biotin
ICOSLG
Conjugated
ICOSLG
IgG1, kappa
25μg
Store at 2-8°C. Do not freeze.
Primary
23308
Videos

For Research Use Only.