missing translation for 'onlineSavingsMsg'
Learn More

CD275 (B7-H2), APC, clone: MIH12, eBioscience™

Mouse Monoclonal Antibody

Brand:  Affymetrix eBioscience 17-5889-41

Code : NEW

Additional Details : Weight : 0.23750kg

 View more versions of this product

Product Code. 15538156

  • £82.61 / 25 tests


EU Stock 5
Add to basket

Description

Description

Description: The MIH12 monoclonal antibody reacts with human B7RP-1, also known as B7h, B7-H2, GL50 and ICOS Ligand. B7RP-1, a member of the B7 family, has a predicted molecular weight of approximately 40kDa and belongs to the Ig superfamily. Human B7RP-1 is expressed by activated monocytes/macrophages. B7RP-1 binds to the ICOS molecule (AILIM, CRP-1) expressed by activated T cells. The interaction of ICOS/B7RP-1 plays an important role in the T cell costimulation pathway.Applications Reported: This MIH12 antibody has been reported for use in flow cytometric analysis.Applications Tested: This MIH12 antibody has been pre-titrated and tested by flow cytometric analysis of normal human peripheral blood monocyte-derived dendritic cells. This can be used at 5 μL (0.125 μg) per test. A test is defined as the amount (μg) of antibody that will stain a cell sample in a final volume of 100 μL. Cell number should be determined empirically but can range from 10^5 to 10^8 cells/test.Excitation: 633-647 nm; Emission: 660 nm; Laser: Red Laser.Filtration: 0.2μm post-manufacturing filtered.
Specifications

Specifications

CD275 (B7-H2)
Monoclonal
5μl/Test
PBS with 0.1% gelatin, 0.2% BSA and 0.09% sodium azide; pH 7.2
O75144
B7-H2, B7H2, B7RP-1, B7RP1, CD275, GL50, ICOS-L, ICOSL, KIAA0653, LICOS, ICOS Ligand
Murine
Affinity chromatography
RUO
Antibody
Monoclonal
Human
Flow Cytometry
MIH12
APC
ICOSLG
Conjugated
ICOSLG
IgG1, kappa
25 Tests
Store at 2-8°C. Do not freeze. Light-sensitive material.
Primary
23308
Videos

For Research Use Only.