missing translation for 'onlineSavingsMsg'
Learn More

CD274 (PD-L1, B7-H1), Biotin, clone: MIH5, eBioscience™

Rat Monoclonal Antibody

Brand:  Affymetrix eBioscience 13-5982-82

Code : Z2

Additional Details : Weight : 0.25000kg

 View more versions of this product

Product Code. 15299969

  • £124.85 / 100µg


EU Stock 5
Add to basket

Disclaimers:

For Research Use Only.


Description

Description

Description: The MIH5 monoclonal antibody reacts with mouse B7-H1, also known as PD-L1. B7-H1, a member of the B7 family, has a predicted molecular weight of approximately 40kDa and belongs to the Ig superfamily. B7-H1 is expressed on a majority of leukocytes including T, B, NK and DC. B7-H1 is a ligand for PD-1. Interaction of PD-1 with either PD-L1 (B7-H1) or PD-L2 (B7-DC) results in inhibition of T and B cell responses. MIH5 is reported to be a blocking antibody.Applications Reported: The MIH5 antibody has been reported for use in flow cytometric analysis.Applications Tested: The MIH5 antibody has been tested by flow cytometric analysis of mouse splenocytes. This can be used at less than or equal to 0.25 μg per test. A test is defined as the amount (μg) of antibody that will stain a cell sample in a final volume of 100 μL. Cell number should be determined empirically but can range from 10^5 to 10^8 cells/test. It is recommended that the antibody be carefully titrated for optimal performance in the assay of interest.Filtration: 0.2μm post-manufacturing filtered.
Specifications

Specifications

CD274 (B7-H1)
MIH5
Biotin
Conjugated
Rat
100μg
Store at 2-8°C. Do not freeze.
Monoclonal
Flow Cytometry
0.5mg/mL
aqueous buffer, 0.09% sodium azide, may contain carrier protein/stabilizer
B7H1, PD-L1, PDL-1
IgG2a
RUO
Primary
Murine
Videos