missing translation for 'onlineSavingsMsg'
Learn More

CD274 (PD-L1, B7-H1), Biotin, clone: MIH1, eBioscience™

Mouse Monoclonal Antibody

Brand:  Affymetrix eBioscience 13-5983-82

Code : Z2

Additional Details : Weight : 0.25000kg

 View more versions of this product

Product Code. 15229979

  • £137.64 / 100µg


Estimated Shipment date
from Supplier 03-07-2018
Add to basket

Description

Description

Description: The MIH1 monoclonal antibody reacts with human B7-H1, also known as PD-L1. B7-H1, a member of the B7 family, has a predicted molecular weight of approximately 40kDa and belongs to the Ig superfamily. B7-H1 is expressed on a majority of leukocytes. B7-H1 is a ligand for PD-1. Interaction of PD-1 with either PD-L1 (B7-H1) or PD-L2 (B7-DC) results in inhibition of T and B cell responses. MIH1 is reported to be a blocking antibody.Applications Reported: The MIH1 antibody has been reported for use in flow cytometric analysis.Applications Tested: The MIH1 antibody has been tested by flow cytometric analysis of normal human peripheral blood cells. This can be used at less than or equal to 0.5 μg per test. A test is defined as the amount (μg) of antibody that will stain a cell sample in a final volume of 100 μL. Cell number should be determined empirically but can range from 10^5 to 10^8 cells/test. It is recommended that the antibody be carefully titrated for optimal performance in the assay of interest.Filtration: 0.2μm post-manufacturing filtered.
Specifications

Specifications

CD274 (B7-H1)
Monoclonal
0.5mg/mL
PBS with 0.1% gelatin and 0.09% sodium azide; pH 7.2
Q9NZQ7
B7-H, B7H1, PD-L1, PDCD1L1, PDCD1LG1, PDL1, B7 homolog 1, CD274 antigen, PDCD1 ligand 1, programmed cell death 1 ligand 1, programmed death ligand 1
Murine
Affinity chromatography
RUO
Antibody
Monoclonal
Human
Flow Cytometry
MIH1
Biotin
CD274
Conjugated
CD274
IgG1, kappa
100μg
Store at 2-8°C. Do not freeze.
Primary
29126
Videos

For Research Use Only.