missing translation for 'onlineSavingsMsg'
Learn More

CD273 (B7-DC), PE, clone: TY25, eBioscience™

Rat Monoclonal Antibody

Brand:  Affymetrix eBioscience 12-5986-83

Code : Z2

Additional Details : Weight : 0.25000kg

 View more versions of this product

Product Code. 15279229

  • £199.23 / 200µg


Estimated Shipment date
from Supplier 29-05-2018
Add to basket

Description

Description

Description: The TY25 monoclonal antibody reacts with mouse B7-DC/PD-L2. B7-DC, a member of the B7 family, has a predicted molecular weight of ∽25kDa and belongs to the Ig superfamily. The mouse B7-DC has a short cytoplasmic tail (4aa). B7-DC is primarily expressed by sub-populations of dendritic cells, monocytes and macrophages. Although B7-DC has structural and sequence similarities to the B7 family, it does not bind CD28/CTLA-4, but binds PD-1. The interactions between PD-1 and B7-DC/PD-L2 have been reported to be involved in costimulation or suppression of T cell proliferation depending on state of cellular activation. TY25 is a useful tool to study the exact function of B7-DC/PD-L2 in APC/T cell interaction and to characterize the expression pattern of this molecule in mouse.Applications Reported: The TY25 antibody has been reported for use in flow cytometric analysis.Applications Tested: The TY25 antibody has been tested by flow cytometric analysis of mouse splenocytes and B7-DC transfected cells. This can be used at less than or equal to 0.5 μg per test. A test is defined as the amount (μg) of antibody that will stain a cell sample in a final volume of 100 μL. Cell number should be determined empirically but can range from 10^5 to 10^8 cells/test. It is recommended that the antibody be carefully titrated for optimal performance in the assay of interest.Excitation: 488-561 nm; Emission: 578 nm; Laser: Blue Laser, Green Laser, Yellow-Green Laser.Filtration: 0.2μm post-manufacturing filtered.
Specifications

Specifications

CD273 (B7-DC)
Monoclonal
0.2mg/mL
PBS with 0.1% gelatin and 0.09% sodium azide; pH 7.2
Q9WUL5
B7-DC, Btdc, F730015O22Rik, MGC124039, MGC124040, PD-L2
Rat
Affinity chromatography
RUO
Antibody
Monoclonal
Murine
Flow Cytometry
TY25
PE
Pdcd1lg2
Conjugated
Pdcd1lg2
IgG2a, kappa
200μg
Store at 2-8°C. Do not freeze. Light-sensitive material.
Primary
58205
Videos

For Research Use Only.