missing translation for 'onlineSavingsMsg'
Learn More

CD273 (B7-DC), PE, clone: 122, eBioscience™

Rat Monoclonal Antibody

Brand:  Affymetrix eBioscience 12-9972-81

Code : Z2

Additional Details : Weight : 0.25000kg

 View more versions of this product

Product Code. 15289369

  • £81.11 / 50µg


Estimated Shipment date
from Supplier 31-05-2018
Add to basket

Description

Description

Description: The 122 monoclonal antibody reacts with mouse B7-DC, also known as PD-L2. B7-DC, a recently identified member of the B7 family, has a predicted molecular weight of approximately 25kDa and belongs to the Ig superfamily. The mouse B7-DC has a short cytoplasmic tail (4aa). B7-DC is primarily expressed by subpopulations of dendritic cells and monocytes/macrophages in the mouse. Although B7-DC has structural and sequence similarities to the B7 family, it does not bind CD28/CTLA-4, rather it is a ligand for PD-1. The interactions between PD-1 and B7-DC/PD-L2 have been reported to be involved in costimulation or suppression of T cell proliferation depending on state of cellular activation. 122 has been demonstrated to block binding of TY25 (Cat. No.14-5986), another mAb specific for mouse B7-DC.Applications Reported: 122 has been reported for use in flow cytometric analysis.Applications Tested: The 122 antibody has been tested by flow cytometric analysis of mouse dendritic cells or mouse B7-DC transfected cells. This can be used at less than or equal to 0.125 μg per test. A test is defined as the amount (μg) of antibody that will stain a cell sample in a final volume of 100 μL. Cell number should be determined empirically but can range from 10^5 to 10^8 cells/test. It is recommended that the antibody be carefully titrated for optimal performance in the assay of interest.Excitation: 488-561 nm; Emission: 578 nm; Laser: Blue Laser, Green Laser, Yellow-Green Laser.Filtration: 0.2μm post-manufacturing filtered.
Specifications

Specifications

CD273 (B7-DC)
Monoclonal
0.2mg/mL
PBS with 0.1% gelatin and 0.09% sodium azide; pH 7.2
Q9WUL5
B7-DC, Btdc, F730015O22Rik, MGC124039, MGC124040, PD-L2
Rat
Affinity chromatography
RUO
Antibody
Monoclonal
Murine
Flow Cytometry
122
PE
Pdcd1lg2
Conjugated
Pdcd1lg2
IgG2a, kappa
50μg
Store at 2-8°C. Do not freeze. Light-sensitive material.
Primary
58205
Videos

For Research Use Only.