Learn More
CD273 (B7-DC) Monoclonal Antibody (MIH18), PerCP-eFluor™ 710, eBioscience™, Invitrogen™
Mouse Monoclonal Antibody
Brand: Affymetrix eBioscience 46-5888-42
Code : NEW
Additional Details : Weight : 0.23750kg
Description
Description: The MIH18 monoclonal antibody reacts with human CD273, also known as PD-L2 or B7-DC, a member of the B7 family that has a predicted molecular weight of approximately 25 kDa, and belongs to the Ig superfamily. B7-DC is primarily expressed by subpopulations of dendritic cells and monocytes/macrophages. Although B7-DC has structural and sequence similarities to the B7 family, it does not bind CD28/CTLA-4, rather it is a ligand for PD-1. The interactions between PD-1 and B7-DC/PD-L2 have been reported to be involved in costimulation or suppression of T cell proliferation depending on state of cellular activation. MIH18 is a useful tool for studies to elucidate the exact function of B7-DC/PD-L2 in APC/T cell interaction, and to characterize the expression pattern of this molecule in humans. MIH18 is a blocking antibody. Applications Reported: This MIH18 antibody has been reported for use in flow cytometric analysis. Applications Tested: This MIH18 antibody has been pre-titrated and tested by flow cytometric analysis of human monocyte-derived dendritic cells. This can be used at 5 μL (0.06 μg) per test. A test is defined as the amount (μg) of antibody that will stain a cell sample in a final volume of 100 μL. Cell number should be determined empirically but can range from 10^5 to 10^8 cells/test.
CD273 (B7-DC) Monoclonal antibody specifically detects CD273 (B7-DC) in Human samples. It is validated for Flow Cytometry.Specifications
CD273 (B7-DC) | |
Monoclonal | |
5 μL/Test | |
PBS with 0.2% BSA and 0.09% sodium azide; pH 7.2 | |
Q9BQ51 | |
PDCD1LG2 | |
Affinity chromatography | |
RUO | |
80380 | |
4° C, store in dark, DO NOT FREEZE! | |
Liquid |
Flow Cytometry | |
MIH18 | |
PerCP-eFluor 710 | |
PDCD1LG2 | |
B7 dendritic cell molecule; B7DC; B7-DC; bA574F11.2; Btdc; Butyrophilin B7-DC; butyrophilin-like protein; CD273; F730015O22Rik; MGC124039; MGC124040; PD1 ligand 2; PD-1 ligand 2; PD-1-ligand 2; PDCD1 ligand 2; PDCD1L2; Pdcd1lg2; Pdl2; PD-L2; Programmed cell death 1 ligand 2; programmed death ligand 2 | |
Mouse | |
100 Tests | |
Primary | |
Human | |
Antibody | |
IgG1 κ |
For Research Use Only.