missing translation for 'onlineSavingsMsg'
Learn More

CD273 (B7-DC), APC, clone: MIH18, eBioscience™

Mouse Monoclonal Antibody

Brand:  Affymetrix eBioscience 17-5888-41

Code : NEW

Additional Details : Weight : 0.23750kg

 View more versions of this product

Product Code. 15518156

  • £74.54 / 25 tests


Estimated Shipment date
from Supplier 29-06-2018
Add to basket

Description

Description

Description: The MIH18 monoclonal antibody reacts with human CD273, also known as PD-L2 or B7-DC, a member of the B7 family that has a predicted molecular weight of approximately 25 kDa, and belongs to the Ig superfamily. B7-DC is primarily expressed by subpopulations of dendritic cells and monocytes/macrophages. Although B7-DC has structural and sequence similarities to the B7 family, it does not bind CD28/CTLA-4, rather it is a ligand for PD-1. The interactions between PD-1 and B7-DC/PD-L2 have been reported to be involved in costimulation or suppression of T cell proliferation depending on state of cellular activation. MIH18 is a useful tool for studies to elucidate the exact function of B7-DC/PD-L2 in APC/T cell interaction, and to characterize the expression pattern of this molecule in humans. MIH18 is a blocking antibody.Applications Reported: This MIH18 antibody has been reported for use in flow cytometric analysis.Applications Tested: This MIH18 antibody has been pre-titrated and tested by flow cytometric analysis of human monocyte-derived dendritic cells. This can be used at 5 μL (0.125 μg) per test. A test is defined as the amount (μg) of antibody that will stain a cell sample in a final volume of 100 μL. Cell number should be determined empirically but can range from 10^5 to 10^8 cells/test.Excitation: 633-647 nm; Emission: 660 nm; Laser: Red Laser.Filtration: 0.2μm post-manufacturing filtered.
Specifications

Specifications

CD273 (B7-DC)
Monoclonal
5μl/Test
PBS with 0.1% gelatin, 0.2% BSA and 0.09% sodium azide; pH 7.2
Q9BQ51
B7-DC, Btdc, F730015O22Rik, MGC124039, MGC124040, PD-L2
Murine
Affinity chromatography
RUO
Antibody
Monoclonal
Human
Flow Cytometry
MIH18
APC
PDCD1LG2
Conjugated
PDCD1LG2
IgG1, kappa
25 Tests
Store at 2-8°C. Do not freeze. Light-sensitive material.
Primary
80380
Videos

For Research Use Only.