missing translation for 'onlineSavingsMsg'
Learn More

CD27, Biotin, clone: O323, eBioscience™

Mouse Monoclonal Antibody

Brand:  Affymetrix eBioscience 13-0279-80

Code : Z2

Additional Details : Weight : 0.25000kg

 View more versions of this product

Product Code. 15279509

  • £49.41 / 25µg


Estimated Shipment date
from Supplier 27-07-2018
Add to basket

Description

Description

Description: The O323 monoclonal antibody reacts with human CD27, a lymphocyte-specific member of the TNFR superfamily. CD27 is expressed by a subset of thymocytes and virtually all mature T cells and is upregulated upon T-cell stimulation. CD27 binds to CD70, and through this interaction, plays an important role in T cell-B cell interaction.Applications Reported: The O323 antibody has been reported for use in flow cytometric analysis.Applications Tested: The O323 antibody has been tested by flow cytometric analysis of normal human peripheral blood cells. This can be used at less than or equal to 1 μg per test. A test is defined as the amount (μg) of antibody that will stain a cell sample in a final volume of 100 μL. Cell number should be determined empirically but can range from 10^5 to 10^8 cells/test. It is recommended that the antibody be carefully titrated for optimal performance in the assay of interest.Filtration: 0.2μm post-manufacturing filtered.
Specifications

Specifications

CD27
Monoclonal
0.5mg/mL
PBS with 0.1% gelatin and 0.09% sodium azide; pH 7.2
P26842
CD27 antigen, CD27L receptor, T cell activation antigen CD27, T cell activation antigen S152, T-cell activation antigen CD27, tumor necrosis factor receptor superfamily member 7
Murine
Affinity chromatography
RUO
Antibody
Monoclonal
Human
Flow Cytometry
O323
Biotin
CD27
Conjugated
CD27
IgG1, kappa
25μg
Store at 2-8°C. Do not freeze.
Primary
939
Videos

For Research Use Only.