missing translation for 'onlineSavingsMsg'
Learn More

CD27, PE-Cyanine7, clone: LG.7F9, eBioscience™

Armenian Hamster Monoclonal Antibody

Brand:  Affymetrix eBioscience 25-0271-82

Code : Z2

Additional Details : Weight : 0.25000kg

 View more versions of this product

Product Code. 15350910

  • £205.37 / 100µg


EU Stock 6
Add to basket

Description

Description

Description: The LG.7F9 monoclonal antibody reacts with mouse CD27, a lymphocyte-specific member of the TNFR superfamily. CD27 is expressed by virtually all mature T cells and by a subpopulation of B cells, mainly memory B cells. In mouse, CD27 has been found on nearly all thymocytes excluding a population of CD46^-CD8^- precursors. CD27 binds to CD70 and, through this interaction, plays an important role in T cell-B cell interaction. It has been reported that triggering CD27 plays an important role in the maturation of CD4^+ and CD8^+ effector cells. LG.7F9 cross-reacts with human and rat CD27.Applications Reported: This LG.7F9 antibody has been reported for use in flow cytometric analysis.Applications Tested: This LG.7F9 antibody has been tested by flow cytometric analysis of mouse splenocytes. This can be used at less than or equal to 0.5 μg per test. A test is defined as the amount (μg) of antibody that will stain a cell sample in a final volume of 100 μL. Cell number should be determined empirically but can range from 10^5 to 10^8 cells/test. It is recommended that the antibody be carefully titrated for optimal performance in the assay of interest.Light sensitivity: This tandem dye is sensitive photo-induced oxidation. Please protect this vial and stained samples from light. Fixation: Samples can be stored in IC Fixation Buffer (cat. 00-8222) (100 μL cell sample + 100 μL IC Fixation Buffer) or 1-step Fix/Lyse Solution (cat. 00-5333) for up to 3 days in the dark at 4°C with minimal impact on brightness and FRET efficiency/compensation. Some generalizations regarding fluorophore performance after fixation can be made, but clone specific performance should be determined empirically.Excitation: 488-561 nm; Emission: 775 nm; Laser: Blue Laser, Green Laser, Yellow-Green Laser.Filtration: 0.2μm post-manufacturing filtered.
Specifications

Specifications

CD27
Monoclonal
0.2mg/mL
PBS with 0.1% gelatin and 0.09% sodium azide; pH 7.2
P26842, P41272
CD27 antigen, CD27L receptor, T cell activation antigen CD27, T cell activation antigen S152, T-cell activation antigen CD27, tumor necrosis factor receptor superfamily member 7
Armenian Hamster
Affinity chromatography
RUO
Antibody
Monoclonal
Rat
Flow Cytometry
LG.7F9
PE-Cyanine7
CD27
Conjugated
Cd27
IgG
100μg
Store at 2-8°C. Do not freeze. Light-sensitive material. This tandem dye is sensitive to photo-induced oxidation. Protect this vial from light during storage, handling and experimental procedures.
Primary
939
Videos

For Research Use Only.