missing translation for 'onlineSavingsMsg'
Learn More

CD27, FITC, clone: LG.7F9, eBioscience™

Armenian Hamster Monoclonal Antibody

Brand:  Affymetrix eBioscience 11-0271-82

Code : Z2

Additional Details : Weight : 0.25000kg

 View more versions of this product

Product Code. 15278059

  • £70.03 / 100µg


EU Stock 5
Add to basket

Description

Description

The LG.7F9 monoclonal antibody reacts with mouse CD27, a lymphocyte-specific member of the TNFR superfamily. CD27 is expressed by virtually all mature T cells and by a subpopulation of B cells, mainly memory B cells. In mouse, CD27 has been found on nearly all thymocytes excluding a population of CD46-CD8- precursors. CD27 binds to CD70 and, through this interaction, plays an important role in T cell-B cell interaction. It has been reported that triggering CD27 plays an important role in the maturation of CD4+ and CD8+ effector cells. LG.7F9 cross-reacts with human and rat CD27.
Specifications

Specifications

CD27
LG.7F9
FITC
Conjugated
Armenian Hamster
100μg
Store at 2-8°C. Do not freeze. Light-sensitive material.
Monoclonal
Flow Cytometry
0.5mg/mL
aqueous buffer, 0.09% sodium azide, may contain carrier protein/stabilizer
TNFRSF7
IgG
RUO
Primary
Rat
Videos

For Research Use Only.