missing translation for 'onlineSavingsMsg'
Learn More

CD268 (BAFF Receptor), APC, clone: eBio7H22-E16, eBioscience™

Rat Monoclonal Antibody

Brand:  Affymetrix eBioscience 17-5943-80

Code : Z2

Additional Details : Weight : 0.25000kg

 View more versions of this product

Product Code. 15300800

  • £81.33 / 25µg


Call for latest availability
+44 (0)1509 555 500
Add to basket

Description

Description

Description: The monoclonal antibody eBio7H22.E16 reacts with mouse BAFF-Receptor, a type III transmembrane TNF receptor family member. BAFF-R was found to be the predominant BAFF receptor expressed on peripheral B cells, in both humans and mice. The receptor is found on the surface of all B220+ B cells found in spleen. BAFF-R is expressed on activated/memory subsets of T-cells and is important for splenic B cell maturation and survival and is a major mediator of BAFF- dependent costimulatory responses in peripheral B and T cells. BAFF-R can cause a high level of autoimmune disease and a defect in BAFF-R will cause a decrease in generating mature B-Cells The ligand for the BAFF-R is BAFF also known as BLyS.Applications Reported: This eBio7H22-E16 antibody has been reported for use in flow cytometric analysis.Applications Tested: This eBio7H22-E16 antibody has been tested by flow cytomtric analysis of mouse splenocytes. This can be used at less than or equal to 0.25 μg per test. A test is defined as the amount (μg) of antibody that will stain a cell sample in a final volume of 100 μL. Cell number should be determined empirically but can range from 10^5 to 10^8 cells/test. It is recommended that the antibody be carefully titrated for optimal performance in the assay of interest.Excitation: 633-647 nm; Emission: 660 nm; Laser: Red Laser.Filtration: 0.2μm post-manufacturing filtered.
Specifications

Specifications

CD268 (BAFF Receptor)
eBio7H22-E16
APC
Conjugated
Rat
25μg
Store at 2-8°C. Do not freeze. Light-sensitive material.
Monoclonal
Flow Cytometry
0.2mg/mL
aqueous buffer, 0.09% sodium azide, may contain carrier protein/stabilizer
BAFFR, BAFF-R, TNFRSF13C, BCMD, BR3, BlySR3
IgG1, kappa
RUO
Primary
Murine
Videos

For Research Use Only.