missing translation for 'onlineSavingsMsg'
Learn More

CD268 (BAFF Receptor) Mouse anti-Human, PE, Clone: 8A7, eBioscience™

Mouse Monoclonal Antibody

Brand:  Affymetrix eBioscience 12-9117-41

Code : NEW

Additional Details : Weight : 0.23750kg

 View more versions of this product

Product Code. 15577176

  • £84.50 / 25 tests

Estimated Shipment
29-09-2020

Please to purchase this item. Need a web account? Register with us today!


Description

Description

The protein encoded by this gene is a cytokine that belongs to the tumor necrosis factor (TNF) ligand family. This cytokine is a ligand for receptors TNFRSF13B/TACI, TNFRSF17/BCMA, and TNFRSF13C/BAFFR. This cytokine is expressed in B cell lineage cells, and acts as a potent B cell activator. It has been also shown to play an important role in the proliferation and differentiation of B cells.
Specifications

Specifications

CD268 (BAFF Receptor)
Monoclonal
5 μL/Test
PBS with 0.1% gelatin, 0.2% BSA and 0.09% sodium azide; pH 7.2
Q96RJ3
TNFRSF13C, BAFF-R, Tumor necrosis factor receptor superfamily member 13C, B-cell-activating factor receptor, BAFF-R, BLyS receptor 3, TNFRSF13C, BAFFR, BR3
Mouse
Affinity chromatography
RUO
Antibody
Monoclonal
Human
Flow Cytometry
8A7
PE
TNFRSF13C
Liquid
TNFRSF13C
IgG2a, kappa
25 Tests
4° C, store in dark, DO NOT FREEZE!
Primary
115650
Videos
Documents

For Research Use Only.