missing translation for 'onlineSavingsMsg'
Learn More

CD268 (BAFF Receptor), FITC, clone: 8A7, eBioscience™

Mouse Monoclonal Antibody

Brand:  Affymetrix eBioscience 11-9117-41

Code : NEW

Additional Details : Weight : 0.23750kg

 View more versions of this product

Product Code. 15526616

  • £84.97 / 25 tests


Estimated Shipment date
from Supplier 27-06-2018
Add to basket

Description

Description

Description: The 8A7 monoclonal antibody reacts with the human BAFF receptor, also known as B lymphocyte stimulator (BLyS protein) receptor, which is expressed on B cells. The ligand for this receptor, BAFF (B cell-activating factor of the TNF family) is a B cell survival factor and regulates CD21/35 and CD23 expression. Interaction of this ligand with its receptor causes elevated CD21/35 and CD23 expression, whereas receptor blockade has been shown to down-modulate expression.Applications Reported: The 8A7 antibody has been reported for use in flow cytometric analysis.Applications Tested: The 8A7 antibody has been pre-titrated and tested by flow cytometric analysis of normal human peripheral blood cells. This can be used at 5 μL (0.125 μg) per test. A test is defined as the amount (μg) of antibody that will stain a cell sample in a final volume of 100 μL. Cell number should be determined empirically but can range from 10^5 to 10^8 cells/test.Excitation: 488 nm; Emission: 520 nm; Laser: Blue Laser.Filtration: 0.2μm post-manufacturing filtered.
Specifications

Specifications

CD268 (BAFF Receptor)
Monoclonal
5μl/Test
PBS with 0.1% gelatin, 0.2% BSA and 0.09% sodium azide; pH 7.2
Q96RJ3
TNFRSF13C, BAFF-R, Tumor necrosis factor receptor superfamily member 13C, B-cell-activating factor receptor, BAFF-R, BLyS receptor 3, TNFRSF13C, BAFFR, BR3
Murine
Affinity chromatography
RUO
Antibody
Monoclonal
Human
Flow Cytometry
8A7
FITC
TNFRSF13C
Conjugated
TNFRSF13C
IgG2a, kappa
25 Tests
Store at 2-8°C. Do not freeze. Light-sensitive material.
Primary
115650
Videos

For Research Use Only.