missing translation for 'onlineSavingsMsg'
Learn More

CD267 (TACI), PE, clone: 11H3, eBioscience™

Mouse Monoclonal Antibody

Brand:  Affymetrix eBioscience 12-9217-41

Code : NEW

Additional Details : Weight : 0.23750kg

 View more versions of this product

Product Code. 15517196

  • £94.27 / 25 tests


EU Stock 1
Add to basket

Disclaimers:

For Research Use Only.


Description

Description

Description: The 11H3 antibody reacts with human TACI, also known as tumor necrosis factor receptor 13B. TACI, like BCMA, binds BAFF and April. TACI and its ligands regulate humoral immune responses, activate NF-kB and c-jun N-terminal kinase, and are involved in B cell associated autoimmune diseases. TACI mRNA and protein are found predominantly in B-cells and correlate with BLyS binding in a panel of B-cell lines. TACI interacts with nanomolar affinity with the BLyS-related tumor necrosis factor homologue APRIL for which no clear in vivo role has been described. 11H3 does not block binding of BAFF/BLyS to TACI.Applications Reported: This 11H3 antibody has been reported for use in flow cytometric analysis.Applications Tested: This 11H3 antibody has been pre-titrated and tested by flow cytometric analysis of normal human peripheral blood cells. This can be used at 5 μL (0.25 μg) per test. A test is defined as the amount (μg) of antibody that will stain a cell sample in a final volume of 100 μL. Cell number should be determined empirically but can range from 10^5 to 10^8 cells/test.Excitation: 488-561 nm; Emission: 578 nm; Laser: Blue Laser, Green Laser, Yellow-Green Laser.Filtration: 0.2μm post-manufacturing filtered.
Specifications

Specifications

CD267 (TACI)
11H3
PE
Conjugated
Murine
25 Tests
Store at 2-8°C. Do not freeze. Light-sensitive material.
Monoclonal
Flow Cytometry
5μL (0.25μg)/test
aqueous buffer, 0.09% sodium azide, may contain carrier protein/stabilizer
TNFRSF13B
IgG2a
RUO
Primary
Human
Videos