missing translation for 'onlineSavingsMsg'
Learn More

CD265 (RANK), PE, clone: R12-31, eBioscience™

Rat Monoclonal Antibody

Brand:  Affymetrix eBioscience 12-6612-81

Code : Z2

Additional Details : Weight : 0.25000kg

 View more versions of this product

Product Code. 15219259

  • £81.08 / 50µg


EU Stock 1
Add to basket

Disclaimers:

For Research Use Only.


Description

Description

Description: The R12-31 monoclonal antibody reacts with mouse RANK (Receptor activator of NF-kappaB), a recently cloned member of the TNFR superfamily with no significant homology to other members of this family. RANK ligand (RANKL/TRANCE/ OPGL) binds to RANK on dendritic cells, upregulates the expression of anti-apoptotic protein BcL-XL suggesting a role in dendritic cell survival. The cytoplasmic domain of RANK interacts with TRAF2, TRAF5 and TRAF6. Overexpression of RANK activates NF-kappaB and c-Jun-terminal kinase (JNK) pathways. Recent studies have shown that RANK interaction with TRAF6 activates NF-kappaB, whereas JNK activation is mediated through binding of RANK to TRAF2.Applications Reported: The R12-31 antibody has been reported for use in flow cytometric analysis.Applications Tested: This R12-31 antibody has been tested by flow cytometric analysis of mouse RANK-transfected cells. This can be used at less than or equal to 1 μg per test. A test is defined as the amount (μg) of antibody that will stain a cell sample in a final volume of 100 μL. Cell number should be determined empirically but can range from 10^5 to 10^8 cells/test. It is recommended that the antibody be carefully titrated for optimal performance in the assay of interest.Excitation: 488-561 nm; Emission: 578 nm; Laser: Blue Laser, Green Laser, Yellow-Green Laser.Filtration: 0.2μm post-manufacturing filtered.
Specifications

Specifications

CD265 (RANK)
R12-31
PE
Conjugated
Rat
50μg
Store at 2-8°C. Do not freeze. Light-sensitive material.
Monoclonal
Flow Cytometry
0.2mg/mL
aqueous buffer, 0.09% sodium azide, may contain carrier protein/stabilizer
Receptor Activator of NF-kB
IgG2a
RUO
Primary
Murine
Videos