missing translation for 'onlineSavingsMsg'
Learn More

CD265 (RANK), Biotin, clone: R12-31, eBioscience™

Rat Monoclonal Antibody

Brand:  Affymetrix eBioscience 13-6612-81

Code : Z2

Additional Details : Weight : 0.25000kg

 View more versions of this product

Product Code. 15370000

This item has been discontinued and is no longer available. View the product for possible alternatives or contact our Product Support team on 01509 555888 for assistance
View alternative products »

Disclaimers:

For Research Use Only.


Description

Description

The R12-31 monoclonal antibody reacts with mouse RANK (Receptor activator of NF-κB), a recently cloned member of the TNFR superfamily with no significant homology to other members of this family. RANK ligand (RANKL/TRANCE/ OPGL) binds to RANK on dendritic cells, upregulates the expression of anti-apoptotic protein BcL-XL suggesting a role in dendritic cell survival. The cytoplasmic domain of RANK interacts with TRAF2, TRAF5 and TRAF6. Overexpression of RANK activates NF-κB and c-Jun-terminal kinase (JNK) pathways. Recent studies have shown that RANK interaction with TRAF6 activates NF-κB, whereas JNK activation is mediated through binding of RANK to TRAF2.
Specifications

Specifications

CD265 (RANK)
R12-31
Biotin
Conjugated
Rat
50μg
Store at 2-8°C. Do not freeze.
Monoclonal
Flow Cytometry
0.5mg/mL
aqueous buffer, 0.09% sodium azide, may contain carrier protein/stabilizer
Receptor Activator of NF-kB
IgG2a
RUO
Primary
Murine
Videos