missing translation for 'onlineSavingsMsg'
Learn More

CD262 (DR5), clone: DJR2-4 (7-8), eBioscience™

Mouse Monoclonal Antibody

Brand:  Affymetrix eBioscience 14-9908-80

Code : Z2

Additional Details : Weight : 0.01000kg

 View more versions of this product

Product Code. 15248227

  • £62.12 / 25µg


Estimated Shipment
01-03-2018
Add to basket

Disclaimers:

For Research Use Only.


Description

Description

Description: The DJR2-4 monoclonal antibody reacts with human DR5, also known as TRAIL-R2, Apo2, TRICK2 and KILLER. DR5 binds to TRAIL, activates NF-kappaB, and induces TRAIL-mediated apoptosis. DR5 is expressed broadly by normal tissues as well as several tumor cells.Applications Reported: The DJR2-4 (a.k.a. 7-8) antibody has been reported for use in flow cytometric analysis.Applications Tested: The DJR2-4 (a.k.a. 7-8) antibody has been tested by flow cytometric analysis of human MOLT-4 cell line and human DR5-transfected cells. This can be used at less than or equal to 1μg per test. A test is defined as the amount (μg) of antibody that will stain a cell sample in a final volume of 100μL. Cell number should be determined empirically but can range from 10^5 to 10^8 cells/test. It is recommended that the antibody be carefully titrated for optimal performance in the assay of interest.Purity: Greater than 90%, as determined by SDS-PAGE.Aggregation: Less than 10%, as determined by HPLC.Filtration: 0.2μm post-manufacturing filtered.
Specifications

Specifications

CD262 (DR5)
DJR2-4 (a.k.a. 7-8)
Unconjugated
Purified
Murine
25μg
Store at 2-8°C.
Monoclonal
Flow Cytometry
0.5mg/mL
aqueous buffer, 0.09% sodium azide, may contain carrier protein/stabilizer
Trail-R2
IgG1
RUO
Primary
Human
Videos