missing translation for 'onlineSavingsMsg'
Learn More

CD262 (DR5), Functional Grade, clone: MD5-1, eBioscience™

Armenian Hamster Monoclonal Antibody

Brand:  Affymetrix eBioscience 16-5883-82

Code : Z2

Additional Details : Weight : 0.01000kg

 View more versions of this product

Product Code. 15258677

  • £137.15 / 100µg


Estimated Shipment
29-06-2018
Add to basket

Description

Description

Description: The MD5-1 monoclonal antibody reacts with mouse DR5, also known as TRAIL-R2, Apo2, TRICK2, and KILLER. DR5 binds to TRAIL, activates NF-kappaB, and induces TRAIL-mediated apoptosis. DR5 mRNA is expressed broadly by normal tissue and human antigen is also expressed by some tumor cell lines.Applications Reported: The MD5-1 antibody has been reported for use in flow cytometric analysis. It has also been reported for use in functional assays.Applications Tested: The MD5-1 antibody has been tested by flow cytometric analysis of mouse DR5-transfected cells. This can be used at less than or equal to 0.25 μg per test. A test is defined as the amount (μg) of antibody that will stain a cell sample in a final volume of 100 μL. Cell number should be determined empirically but can range from 10^5 to 10^8 cells/test. It is recommended that the antibody be carefully titrated for optimal performance in the assay of interest.Endotoxin: Less than 0.001 ng/ug antibody as determined by the LAL assay.Storage and handling: Use in a sterile environment.Filtration: 0.2μm post-manufacturing filtered.Purity: Greater than 90%, as determined by SDS-PAGE.Endotoxin Level: Less than 0.001 ng/μg antibody, as determined by LAL assay.Aggregation: Less than 10%, as determined by HPLC.
Specifications

Specifications

CD262 (DR5)
Monoclonal
1mg/mL
PBS with no preservative; pH 7.2
Q9QZM4
CD262, DR5, KILLER, KILLER/DR5, TRAIL-R2, TRAILR2, TRICK2, TRICK2A, TRICK2B, TRICKB, ZTNFR9, Trail receptor 2
Armenian Hamster
Affinity chromatography
RUO
Antibody
Monoclonal
Murine
Functional Assay
MD5-1
Functional Grade
Tnfrsf10b
Purified
Tnfrsf10b
IgG
100μg
Store at 2-8°C.
Primary
21933
Videos

For Research Use Only.