Learn More
CD26 Monoclonal Antibody (H194-112 (H194-112.12.4.4.5)), PerCP-Cyanine5.5, eBioscience™, Invitrogen™
Rat Monoclonal Antibody
Brand: Affymetrix eBioscience 45-0261-82
162.67 GBP valid until 2024-03-29
Use promo code "21615" to get your promotional price.
Code : Z2
Additional Details : Weight : 0.25000kg
Description
Description: The monoclonal antibody H194-112 recognizes mouse CD26, a thymocyte-activating molecule (THAM). CD26 is a surface expressed and soluble ectoenzyme (dipeptidyl peptidase IV) that cleaves dipeptides from the amino terminus of proteins. In particular CD26 has been shown to cleave CXCL12, CCL22, neuropeptide Y and glucagon. CD26 is involved in adhesion, apoptosis, disease regulation, and engraftment. Expression is regulated during thymic development at the messenger RNA level. Additionally, CD26 protein is found on thymocytes, a percentage of resting B cells, stimulated B cells, NK cells and a percentage of resting and activated T cells and other epithelial cell types. On human cells more CD26 protein is found on Th2 cells. Functional CD26 is expressed on Sca-1+c-Kit+lin- cells. This expression has been shown to correlate with mobilization of progenitor cells. Inhibition of CD26 activity through a variety of compounds enhances allogenic stem cell homing and engraftment. This antibody is reported to have activating function. Applications Reported: This H194-112 (H194-112.12.4.4.5) antibody has been reported for use in flow cytometric analysis. Applications Tested: This H194-112 (H194-112.12.4.4.5) antibody has been tested by flow cytometric analysis of mouse splenocytes or thymocytes. This can be used at less than or equal to 1 μg per test.
CD26 Monoclonal antibody specifically detects CD26 in Mouse samples. It is validated for Flow Cytometry.Specifications
CD26 | |
Monoclonal | |
0.2 mg/mL | |
PBS with 0.09% sodium azide; pH 7.2 | |
P28843 | |
DPP4 | |
Affinity chromatography | |
RUO | |
13482 | |
4° C, store in dark, DO NOT FREEZE! | |
Liquid |
Flow Cytometry | |
H194-112 (H194-112.12.4.4.5) | |
PerCP-Cyanine5.5 | |
DPP4 | |
ACT3; Activation molecule 3; ADABP; ADCP2; ADCP-2; ADCP-I; adenosine deaminase complexing protein; Adenosine deaminase complexing protein 2; bile canaliculus domain-specific membrane glycoprotein; CD26; Dipeptidyl peptidase 4; Dipeptidyl peptidase 4 60 kDa soluble form; Dipeptidyl peptidase 4 membrane form; Dipeptidyl peptidase 4 soluble form; Dipeptidyl peptidase IV; Dipeptidyl peptidase IV 60 kDa soluble form; Dipeptidyl peptidase IV membrane form; Dipeptidyl peptidase IV soluble form; dipeptidylpeptidase 4; dipeptidyl-peptidase 4; dipeptidyl-peptidase 4 (CD26, adenosine deaminase complexing protein 2); dipeptidylpeptidase IV (CD26, adenosine deaminase complexing protein 2); DPP IV; DPP4; Dpp-4; DPPIV; DPP-IV; GP110 glycoprotein; I79_016618; serine protease; T-cell activation antigen CD26; THAM; Thymocyte-activating molecule; TP103; WC10 | |
Rat | |
100 μg | |
Primary | |
Mouse | |
Antibody | |
IgG2a κ |
For Research Use Only.