missing translation for 'onlineSavingsMsg'
Learn More

CD26, PerCP-Cyanine5.5, clone: H194-112 (H194-112.12.4.4.5), eBioscience™

Rat Monoclonal Antibody

Brand:  Affymetrix eBioscience 45-0261-80

Code : Z2

Additional Details : Weight : 0.25000kg

 View more versions of this product

Product Code. 15371150

  • £74.43 / 25µg


EU Stock 3
Add to basket

Description

Description

Description: The monoclonal antibody H194-112 recognizes mouse CD26, a thymocyte-activating molecule (THAM). CD26 is a surface expressed and soluble ectoenzyme (dipeptidyl peptidase IV) that cleaves dipeptides from the amino terminus of proteins. In particular CD26 has been shown to cleave CXCL12, CCL22, neuropeptide Y and glucagon. CD26 is involved in adhesion, apoptosis, disease regulation, and engraftment. Expression is regulated during thymic development at the messenger RNA level. Additionally, CD26 protein is found on thymocytes, a percentage of resting B cells, stimulated B cells, NK cells and a percentage of resting and activated T cells and other epithelial cell types. On human cells more CD26 protein is found on Th2 cells. Functional CD26 is expressed on Sca-1+c-Kit+lin- cells. This expression has been shown to correlate with mobilization of progenitor cells. Inhibition of CD26 activity through a variety of compounds enhances allogenic stem cell homing and engraftment. This antibody is reported to have activating function.Applications Reported: This H194-112 (H194-112.12.4.4.5) antibody has been reported for use in flow cytometric analysis.Applications Tested: This H194-112 (H194-112.12.4.4.5) antibody has been tested by flow cytometric analysis of mouse splenocytes or thymocytes. This can be used at less than or equal to 1 μg per test. A test is defined as the amount (μg) of antibody that will stain a cell sample in a final volume of 100 μL. Cell number should be determined empirically but can range from 10^5 to 10^8 cells/test. It is recommended that the antibody be carefully titrated for optimal performance in the assay of interest.Excitation: 488 nm; Emission: 695 nm; Laser: Blue Laser.Filtration: 0.2μm post-manufacturing filtered.
Specifications

Specifications

CD26
Monoclonal
0.2mg/mL
PBS with 0.1% gelatin and 0.09% sodium azide; pH 7.2
P28843
ADABP, ADCP2, CD26, DPP-IV, TP103, ADCP-2, DPP IV, T-cell activation antigen CD26, adenosine deaminase complexing protein 2, dipeptidyl peptidase 4, dipeptidyl peptidase IV, dipeptidylpeptidase 4, dipeptidylpeptidase IV CD26, adenosine deaminase complexing protein 2
Rat
Affinity chromatography
RUO
Antibody
Monoclonal
Murine
Flow Cytometry
H194-112 (H194-112.12.4.4.5)
PerCP-Cyanine5.5
Dpp4
Conjugated
Dpp4
IgG2a, kappa
25μg
Store at 2-8°C. Do not freeze. Light-sensitive material.
Primary
13482
Videos

For Research Use Only.