Learn More
CD26 Monoclonal Antibody (BA5b), APC, Invitrogen™
Mouse Monoclonal Antibody
Brand: Invitrogen CD2605
Code : 50
Additional Details : Weight : 0.50000kg
Description
Allophycocyanin (APC) is a stable and highly soluble phycobiliprotein that provides maximal absorbance and fluorescence without susceptibility to internal or external fluorescence quenching, thus providing exceptional quantum yields and molar extinction coefficients.
CD26 Monoclonal antibody specifically detects CD26 in Human samples. It is validated for Flow Cytometry.Specifications
CD26 | |
Monoclonal | |
APC | |
DPP4 | |
ACT3; Activation molecule 3; ADABP; ADCP2; ADCP-2; ADCP-I; adenosine deaminase complexing protein; Adenosine deaminase complexing protein 2; bile canaliculus domain-specific membrane glycoprotein; CD26; Dipeptidyl peptidase 4; Dipeptidyl peptidase 4 60 kDa soluble form; Dipeptidyl peptidase 4 membrane form; Dipeptidyl peptidase 4 soluble form; Dipeptidyl peptidase IV; Dipeptidyl peptidase IV 60 kDa soluble form; Dipeptidyl peptidase IV membrane form; Dipeptidyl peptidase IV soluble form; dipeptidylpeptidase 4; dipeptidyl-peptidase 4; dipeptidyl-peptidase 4 (CD26, adenosine deaminase complexing protein 2); dipeptidylpeptidase IV (CD26, adenosine deaminase complexing protein 2); DPP IV; DPP4; Dpp-4; DPPIV; DPP-IV; GP110 glycoprotein; I79_016618; serine protease; T-cell activation antigen CD26; THAM; Thymocyte-activating molecule; TP103; WC10 | |
Mouse | |
purified | |
ASR | |
1803 | |
4° C, store in dark | |
Liquid |
Flow Cytometry | |
BA5b | |
PBS with BSA, sucrose and 0.1% sodium azide | |
P27487 | |
DPP4 | |
Human CD26 | |
500 μL | |
Primary | |
Human | |
Antibody | |
IgG2a κ |