missing translation for 'onlineSavingsMsg'
Learn More

CD257 (BAFF, BLyS), PE, clone: 1D6, eBioscience™

Mouse Monoclonal Antibody

Brand:  Affymetrix eBioscience 12-9017-41

Code : NEW

Additional Details : Weight : 0.23750kg

 View more versions of this product

Product Code. 15577156

  • £87.94 / 25 tests


Estimated Shipment date
from Supplier 27-07-2018
Add to basket

Description

Description

Description: The 1D6 monoclonal antibody reacts with human BLyS (B lymphocyte stimulator), also known as BAFF (B cell activating factor from the TNF family), TALL-1 (TNF and apoptosis leukocyte-expressed ligand-1), THANK (TNF homologue that activates apoptosis, nuclear factor KappaB and c-Jun NH2-terminal kinase) and zTNF4. BLyS, a member of the TNF family, is a type II membrane protein that exists in both membrane-bound and soluble forms. Expression of membrane-bound BLyS on monocytes lineage is regulated by IFN gamma. BLyS acts as a potent B cell growth factor and costimulator of Ig production. BLyS transgenic mice develop severe B cell hyperplasia and autoimmune lupus-like disease characterized by the presence of autoantibodies against nuclear antigens and immune complexes deposited in the kidney. Data from animal models imply a role for BLyS in human autoimmune diseases. To date BCMA, TACI and BR3 have been identified as receptors for BLyS. APRIL, another TNF family member, and BLyS can form active heterotrimeric molecules when coexpressed and these circulating heterotrimers are present in serum samples from patients with systemic immune-based rheumatic diseases. Furthermore, it has been reported that BLyS and APRIL, share BCMA and TACI as their receptors.Applications Reported: The 1D6 antibody has been reported for use in flow cytometric analysis.Applications Tested: This 1D6 antibody has been pre-titrated and tested by flow cytometric analysis of human BLyS transfected cells. This can be used at 5 μL (0.5 μg) per test. A test is defined as the amount (μg) of antibody that will stain a cell sample in a final volume of 100 μL. Cell number should be determined empirically but can range from 10^5 to 10^8 cells/test.Excitation: 488-561 nm; Emission: 578 nm; Laser: Blue Laser, Green Laser, Yellow-Green Laser.Filtration: 0.2μm post-manufacturing filtered.
Specifications

Specifications

CD257 (BAFF, BLyS)
Monoclonal
5μl/Test
PBS with 0.1% gelatin, 0.2% BSA and 0.09% sodium azide; pH 7.2
Q9Y275
ApoL related ligand TALL-1, B-cell-activating factor, B-lymphocyte stimulator, BLYS, delta BAFF, Delta4 BAFF, dendritic cell-derived TNF-like molecule, DTL, TALL-1, TALL1, THANK, TNF and ApoL-related leukocyte expressed ligand 1, TNF homolog that activates apoptosis, TNFSF13B, TNFSF20, tumor necrosis factor (ligand) superfamily member 13b, tumor necrosis factor (ligand) superfamily, member 13b, tumor necrosis factor (ligand) superfamily, member 20, tumor necrosis factor ligand 7A, tumor necrosis factor-like protein ZTNF4, ZTNF4
Murine
Affinity chromatography
RUO
Antibody
Monoclonal
Human
Flow Cytometry
1D6
PE
TNFSF13B
Conjugated
TNFSF13B
IgG1, kappa
25 Tests
Store at 2-8°C. Do not freeze. Light-sensitive material.
Primary
10673
Videos

For Research Use Only.