missing translation for 'onlineSavingsMsg'
Learn More

CD254 (RANK Ligand), clone: MIH24, eBioscience™

Mouse Monoclonal Antibody

Brand:  Affymetrix eBioscience 14-6619-80

Code : Z2

Additional Details : Weight : 0.01000kg

 View more versions of this product

Product Code. 15227927

  • £75.12 / 25µg


Call for latest availability
+44 (0)1509 555 500
Add to basket

Description

Description

Description: The MIH24 monoclonal antibody reacts with human TRANCE (TNF-related activation-induced cytokine receptor) also known as RANKL (receptor activator of NF-kappaB ligand) and OPGL (Osteoprotegerin Ligand). TRANCE is a type II membrane protein with predicted molecular weight of 35 kDa. Its extracellular domain is most closely related to TRAIL, FasL and TNF. TRANCE mRNA is upregulated by TCR stimulation. TRANCE-deficient mouse lacks osteoclasts. In addition, TRANCE has been shown to be involved in the interactions between T-dendritic and T-B cells. It was also found to be critical in osteoclast differentiation. The TRANCE-RANK interaction regulates the expression of the anti-apoptotic molecule Bcl-xL.Applications Reported: The MIH24 antibody has been reported for use in flow cytometric analysis, and ELISA.Applications Tested: The MIH24 antibody has been tested by flow cytometric analysis of human RANKL transfected cells. This can be used at less than or equal to 1 μg per test. A test is defined as the amount (μg) of antibody that will stain a cell sample in a final volume of 100 μL. Cell number should be determined empirically but can range from 10^5 to 10^8 cells/test. It is recommended that the antibody be carefully titrated for optimal performance in the assay of interest.Purity: Greater than 90%, as determined by SDS-PAGE.Aggregation: Less than 10%, as determined by HPLC.Filtration: 0.2μm post-manufacturing filtered.
Specifications

Specifications

CD254 (RANK Ligand)
Monoclonal
0.5mg/mL
PBS with 0.09% sodium azide; pH 7.2
O14788
RP11-86N24.2, CD254, ODF, OPGL, OPTB2, RANKL, TRANCE, hRANKL2, sOdf, TNF-related activation-induced cytokine, TNFSF11, TRANCE
Murine
Affinity chromatography
RUO
Antibody
Monoclonal
Human
ELISAþFlow Cytometry
MIH24
Unconjugated
TNFSF11
Purified
TNFSF11
IgG2b, kappa
25μg
Store at 2-8°C.
Primary
8600
Videos

For Research Use Only.