missing translation for 'onlineSavingsMsg'
Learn More

CD253 (TRAIL), Biotin, clone: N2B2, eBioscience™

Rat Monoclonal Antibody

Brand:  Affymetrix eBioscience 13-5951-81

Code : Z2

Additional Details : Weight : 0.25000kg

 View more versions of this product

Product Code. 15299939

  • £68.96 / 50µg


Estimated Shipment date
from Supplier 04-06-2018
Add to basket

Description

Description

Description: The N2B2 monoclonal antibody reacts with mouse TNF-related apoptosis-inducing ligand (TRAIL), a member of the TNF superfamily. TRAIL is not detected on the surface of freshly isolated T, B, or NK cells, but can be induced preferentially on CD3^- NK1.1^+ NK cells after stimulation with IL-2 or IL-15. N2B2 inhibits IL-2- or IL-15-activated NK cell cytotoxicity against mouse fibrosarcoma L929 target cells.Applications Reported: N2B2 has been reported for use in flow cytometric analysis.Applications Tested: The N2B2 antibody has been tested by flow cytometric analysis of mouse TRAIL transfected cells and can be used at less than or equal to 0.25 μg per test. A test is defined as the amount (μg) of antibody that will stain a cell sample in a final volume of 100 μL. Cell number should be determined empirically but can range from 10^5 to 10^8 cells/test. It is recommended that the antibody be carefully titrated for optimal performance in the assay of interest.Filtration: 0.2μm post-manufacturing filtered.
Specifications

Specifications

CD253 (TRAIL)
Monoclonal
0.5mg/mL
PBS with 0.1% gelatin and 0.09% sodium azide; pH 7.2
P50592
Apo2L (for Apo-2 Ligand), TNF-Related Apoptosis-Inducing Ligand, Tumor necrosis factor ligand, superfamily member 10, TRAIL protein, CD253 antigen, TN
Rat
Affinity chromatography
RUO
Antibody
Monoclonal
Murine
Flow Cytometry
N2B2
Biotin
Tnfsf10
Conjugated
Tnfsf10
IgG2a, kappa
50μg
Store at 2-8°C. Do not freeze.
Primary
22035
Videos

For Research Use Only.