missing translation for 'onlineSavingsMsg'
Learn More

CD252 (OX40 Ligand), APC, clone: RM134L, eBioscience™

Rat Monoclonal Antibody

Brand:  Affymetrix eBioscience 17-5905-82

Code : Z2

Additional Details : Weight : 0.25000kg

 View more versions of this product

Product Code. 15350780

  • £143.93 / 100µg


Call for latest availability
+44 (0)1509 555 500
Add to basket

Description

Description

Description: The RM134L monoclonal antibody reacts with mouse CD252 also known as OX-40 Ligand, a member of the TNF superfamily. OX-40L is induced on mouse splenic B cells stimulated with a combination of anti-IgM and anti-CD40. Neither resting nor activated mouse T cells express OX-40L. The interaction of OX-40 (CD134) with OX-40L has been implicated in T cell-dependent humoral response. RM134L inhibits the costimulatory activity of mouse OX-40L transfected cells for anti-CD3-stimulated T cell proliferation.Applications Reported: This RM134L antibody has been reported for use in flow cytometric analysis.Applications Tested: This RM134L antibody has been tested by flow cytometric analysis of stimulated mouse splenocytes. This can be used at less than or equal to 0.5 μg per test. A test is defined as the amount (μg) of antibody that will stain a cell sample in a final volume of 100 μL. Cell number should be determined empirically but can range from 10^5 to 10^8 cells/test. It is recommended that the antibody be carefully titrated for optimal performance in the assay of interest.Excitation: 633-647 nm; Emission: 660 nm; Laser: Red Laser.Filtration: 0.2μm post-manufacturing filtered.
Specifications

Specifications

CD252 (OX40 Ligand)
Monoclonal
0.2mg/mL
PBS with 0.1% gelatin and 0.09% sodium azide; pH 7.2
P43488
Ath-1, Ath1, TXGP1, Txgp1l, gp34, TNFSF4, CD252, OX40 Ligand
Rat
Affinity chromatography
RUO
Antibody
Monoclonal
Murine
Flow Cytometry
RM134L
APC
Tnfsf4
Conjugated
Tnfsf4
IgG2b, kappa
100μg
Store at 2-8°C. Do not freeze. Light-sensitive material.
Primary
22164
Videos

For Research Use Only.