missing translation for 'onlineSavingsMsg'
Learn More

CD25, Biotin, clone: PC61.5, eBioscience™

Rat Monoclonal Antibody

Brand:  Affymetrix eBioscience 13-0251-86

Code : Z2

Additional Details : Weight : 0.25000kg

 View more versions of this product

Product Code. 15289499

  • £249.04 / 1mg


Estimated Shipment date
from Supplier 23-07-2018
Add to basket

Description

Description

Description: The PC61.5 antibody reacts with mouse CD25, the 55kDa interleukin-2 receptor alpha chain (IL-2R alpha). CD25 is expressed by early progenitors of the T and B lineage as well as by activated mature T and B lymphocytes. By itself, CD25 binds IL-2 only with low affinity. However, CD25 associates with CD122 (IL-2 receptor beta chain) and CD132 (common gamma chain) to form the high affinity IL-2 receptor. Binding of IL-2 to both the high and low affinity classes of IL-2 receptor is inhibited by the PC61.5 antibody. CD25 plays a role in lymphocyte differentiation and activation/proliferation.Applications Reported: The PC61.5 antibody has been reported for use in flow cytometric analysis.Applications Tested: The PC61.5 antibody has been tested by flow cytometric analysis of mouse splenocytes and Con A-stimulated splenocytes. This can be used at less than or equal to 0.125 μg per test. A test is defined as the amount (μg) of antibody that will stain a cell sample in a final volume of 100 μL. Cell number should be determined empirically but can range from 10^5 to 10^8 cells/test. It is recommended that the antibody be carefully titrated for optimal performance in the assay of interest.Filtration: 0.2μm post-manufacturing filtered.
Specifications

Specifications

CD25
Monoclonal
0.5mg/mL
PBS with 0.1% gelatin and 0.09% sodium azide; pH 7.2
P01590
IL2 RA antibody, Il2ra antibody, Interleukin 2 receptor alpha chain antibody, TAC antigen antibody, IL-2R alpha chain
Rat
Affinity chromatography
RUO
Antibody
Monoclonal
Murine
Flow Cytometry
PC61.5
Biotin
Il2ra
Conjugated
Il2ra
IgG1, lambda
1mg
Store at 2-8°C. Do not freeze.
Primary
16184
Videos

For Research Use Only.