missing translation for 'onlineSavingsMsg'
Learn More

CD25, Alexa Fluor 700, clone: PC61.5, eBioscience™

Rat Monoclonal Antibody

Brand:  Affymetrix eBioscience 56-0251-82

Code : Z2

Additional Details : Weight : 0.25000kg

 View more versions of this product

Product Code. 15332120

  • £125.97 / 100µg


EU Stock 3
Add to basket

Disclaimers:

For Research Use Only.


Description

Description

Description: The PC61.5 antibody reacts with mouse CD25, the 55kDa interleukin-2 receptor alpha chain (IL-2R alpha). CD25 is expressed by early progenitors of the T and B lineage as well as by activated mature T and B lymphocytes. By itself, CD25 binds IL-2 only with low affinity. However, CD25 associates with CD122 (IL-2 receptor beta chain) and CD132 (common gamma chain) to form the high affinity IL-2 receptor. Binding of IL-2 to both the high and low affinity classes of IL-2 receptor is inhibited by the PC61.5 antibody. CD25 plays a role in lymphocyte differentiation and activation/proliferation.Applications Reported: This PC61.5 antibody has been reported for use in flow cytometric analysis.Applications Tested: This PC61.5 antibody has been tested by flow cytometric analysis of mouse splenocytes. This can be used at less than or equal to 0.25 μg per test. A test is defined as the amount (μg) of antibody that will stain a cell sample in a final volume of 100 μL. Cell number should be determined empirically but can range from 10^5 to 10^8 cells/test. It is recommended that the antibody be carefully titrated for optimal performance in the assay of interest.Alexa Fluor® 700 emits at 723 nm and can be excited with the red laser (633 nm). Most instruments will require a 685 LP mirror and 710/20 filter. Please make sure that your instrument is capable of detecting this fluorochrome.Excitation: 633-647 nm; Emission: 723 nm; Laser: Red Laser.Filtration: 0.2μm post-manufacturing filtered.
Specifications

Specifications

CD25
PC61.5
Alexa Fluor 700
Conjugated
Rat
100μg
Store at 2-8°C. Do not freeze. Light-sensitive material.
Monoclonal
Flow Cytometry
0.2mg/mL
aqueous buffer, 0.09% sodium azide, may contain carrier protein/stabilizer
Interleukin-2 Receptor alpha
IgG1
RUO
Primary
Murine
Videos