missing translation for 'onlineSavingsMsg'
Learn More

CD25, Alexa Fluor 488, clone: eBio3C7 (3C7), eBioscience™

Rat Monoclonal Antibody

Brand:  Affymetrix eBioscience 53-0253-80

Code : Z2

Additional Details : Weight : 0.25000kg

 View more versions of this product

Product Code. 15351990

  • £54.42 / 25µg


EU Stock 4
Add to basket

Disclaimers:

For Research Use Only.


Description

Description

Description: The eBio3C7 antibody reacts with mouse CD25, the 55kDa interleukin-2 receptor a chain (IL-2Ra). CD25 is expressed by early progenitors of the T and B lineage as well as by activated mature T and B lymphocytes. By itself, CD25 binds IL-2 only with low affinity. However, CD25 associates with CD122 (IL-2 receptor beta chain) and CD132 (common gamma chain) to form the high affinity IL-2 receptor. Binding of IL-2 to both the high and low affinity classes of the IL-2 receptor is inhibited by the 3C7 antibody. CD25 plays a role in lymphocyte differentiation and activation/proliferation. The epitopes recognized by eBio3C7, PC61.5 and eBio7D4 are all different allowing for depletion with one antibody (typically PC61.5) and subsequent detection with another CD25 antibody.Applications Reported: This eBio3C7 (3C7) antibody has been reported for use in flow cytometric analysis.Applications Tested: This eBio3C7 (3C7) antibody has been tested by flow cytometric analysis of mouse splenocytes. This can be used at less than or equal to 0.06 μg per test. A test is defined as the amount (μg) of antibody that will stain a cell sample in a final volume of 100 μL. Cell number should be determined empirically but can range from 10^5 to 10^8 cells/test. It is recommended that the antibody be carefully titrated for optimal performance in the assay of interest.Excitation: 488 nm; Emission: 519 nm; Laser: Blue Laser.Filtration: 0.2μm post-manufacturing filtered.
Specifications

Specifications

CD25
eBio3C7 (3C7)
Alexa Fluor 488
Conjugated
Rat
25μg
Store at 2-8°C. Do not freeze. Light-sensitive material.
Monoclonal
Flow Cytometry
0.5mg/mL
aqueous buffer, 0.09% sodium azide, may contain carrier protein/stabilizer
Interleukin-2 Receptor alpha, IL-2Ra, IL2Ra
IgG2b
RUO
Primary
Murine
Videos