missing translation for 'onlineSavingsMsg'
Learn More

CD25, PerCP-Cyanine5.5, clone: BC96, eBioscience™

Mouse Monoclonal Antibody

Brand:  Affymetrix eBioscience 45-0259-42

Code : NEW

Additional Details : Weight : 0.23750kg

 View more versions of this product

Product Code. 15539016

  • £208.21 / 100 tests


EU Stock 17
Add to basket

Description

Description

Description: The BC96 monoclonal antibody reacts with human CD25, the 55kDa interleukin-2 receptor \alpha chain (IL-2R\alpha). CD25 is expressed by early progenitors of T and B lineage as well as by activated mature T and B lymphocytes. By itself, CD25 binds IL-2 only with low affinity. However, CD25 associates with CD122 (IL-2 receptor \beta chain) and CD132 (common \gamma chain) to form the high affinity IL-2 receptor. CD25 plays a role in lymphocyte differentiating and activation/proliferation.Applications Reported: This BC96 antibody has been reported for use in flow cytometric analysis.Applications Tested: This BC96 antibody has been pre-titrated and tested by flow cytometric analysis of normal human peripheral blood cells. This can be used at 5 μL (0.125 μg) per test. A test is defined as the amount (μg) of antibody that will stain a cell sample in a final volume of 100 μL. Cell number should be determined empirically but can range from 10^5 to 10^8 cells/test.Excitation: 488 nm; Emission: 695 nm; Laser: Blue Laser.Filtration: 0.2μm post-manufacturing filtered.
Specifications

Specifications

CD25
Monoclonal
5μl/Test
PBS with 0.1% gelatin, 0.2% BSA and 0.09% sodium azide; pH 7.2
P01589
IL2 RA antibody, Il2ra antibody, Interleukin 2 receptor alpha chain antibody, TAC antigen antibody, IL-2R alpha chain
Murine
Affinity chromatography
RUO
Antibody
Monoclonal
Human
Flow Cytometry
BC96
PerCP-Cyanine5.5
IL2RA
Conjugated
IL2RA
IgG1, kappa
100 Tests
Store at 2-8°C. Do not freeze. Light-sensitive material.
Primary
3559
Videos

For Research Use Only.