missing translation for 'onlineSavingsMsg'
Learn More

CD25 Mouse anti-Human, Non-human primate, PE, Clone: CD25-4E3, eBioscience™

Mouse Monoclonal Antibody

Brand:  Affymetrix eBioscience 12-0257-42

Code : NEW

Additional Details : Weight : 0.23750kg

 View more versions of this product

Product Code. 15596736

  • £148.00 / 100 tests

In stock, ready to ship: 5

Please to purchase this item. Need a web account? Register with us today!


Description

Description

CD25 (IL2 receptor alpha chain/IL2RA) is a cytokine that plays a role in the proliferation of T and B lymphocytes. The receptor of this cytokine (IL2RA) is a heterotrimeric protein complex with a gamma chain also shared by interleukin 4 (IL4) and interleukin 7 (IL7). IL2RA, IL2R beta chain (IL2RB), and the IL2R gamma chain (IL2RG), constitute the high-affinity IL2 receptor. Homodimeric IL2RA chains result in low-affinity receptor, while homodimeric IL2RB chains produce a medium-affinity receptor. The expression of IL2 in mature thymocytes is monoallelic, which represents an unusual regulatory mode for controlling the precise expression of a single gene. IL2 is primarily produced by mature T cells. IL2 plays an important role as a growth factor, differentiation factor, and regulator of cell death. IL-2 stimulates the proliferation of B cells, augments natural killer cell activity, and inhibits granulocyte macrophage colony formation. The targeted disruption of a similar gene in mice leads to ulcerative colitis-like disease, which suggests a role in the immune response to antigenic stimuli. Mutations in this gene are associated with interleukin 2 receptor alpha deficiency.
Specifications

Specifications

CD25
Monoclonal
5 μL/Test
PBS with 0.1% gelatin, 0.2% BSA and 0.09% sodium azide; pH 7.2
P01589
IL2 RA antibody, Il2ra antibody, Interleukin 2 receptor alpha chain antibody, TAC antigen antibody, IL-2R alpha chain
Mouse
Affinity chromatography
RUO
Antibody
Monoclonal
Human, Non-human Primate
Flow Cytometry
CD25-4E3
PE
IL2RA
Liquid
IL2RA
IgG2b, kappa
100 Tests
4° C, store in dark, DO NOT FREEZE!
Primary
3559
Videos
SDS
Documents

For Research Use Only.