missing translation for 'onlineSavingsMsg'
Learn More

CD25, APC, clone: CD25-4E3, eBioscience™

Mouse Monoclonal Antibody

Brand:  Affymetrix eBioscience 17-0257-42

Code : NEW

Additional Details : Weight : 0.23750kg

 View more versions of this product

Product Code. 15587896

  • £174.50 / 100 tests


EU Stock 9
Add to basket

Disclaimers:

For Research Use Only.


Description

Description

Description: This CD25-4E3 monoclonal antibody reacts to human CD25, which is also known as the low-affinity interleukin (IL)-2 receptor alpha. CD25 is expressed on activated T cells (including a subset of regulatory T cells), B cells, and macrophages. This receptor subunit associates with CD122, the IL-2 receptor beta chain, and CD132, the common gamma chain, to form the high-affinity IL-2 receptor. IL-2 receptor signaling has been linked to cell proliferation and survival in lymphocytes. CD25 is a marker for Hodgkin's disease, and has been associated with such autoimmune diseases as Type I diabetes.This monoclonal antibody is cross-reactive to rhesus monkey.Applications Reported: This CD25-4E3 antibody has been reported for use in flow cytometric analysis.Applications Tested: This CD25-4E3 antibody has been pre-titrated and tested by flow cytometric analysis of nrmal human peripheral blood cells. This can be used at 5 μL (0.06 μg) per test. A test is defined as the amount (μg) of antibody that will stain a cell sample in a final volume of 100 μL. Cell number should be determined empirically but can range from 10^5 to 10^8 cells/test.Excitation: 633-647 nm; Emission: 660 nm; Laser: Red Laser.Filtration: 0.2μm post-manufacturing filtered.
Specifications

Specifications

CD25
CD25-4E3
APC
Conjugated
Murine
100 Tests
Store at 2-8°C. Do not freeze. Light-sensitive material.
Monoclonal
Flow Cytometry
5μL (0.06μg)/test
aqueous buffer, 0.09% sodium azide, may contain carrier protein/stabilizer
Interleukin-2 Receptor alpha, IL-2Ra
IgG2b
RUO
Primary
Non-human Primate
Videos