missing translation for 'onlineSavingsMsg'
Learn More

CD25, clone: GLLZDMY, eBioscience™

Mouse Monoclonal Antibody

Brand:  Affymetrix eBioscience 14-0256-82

Code : NEW

Additional Details : Weight : 0.09500kg

Product Code. 15577416

  • £119.00 / 100µg

Estimated Shipment
29-10-2020

Please to purchase this item. Need a web account? Register with us today!


Description

Description

Description: This GLLZDMY monoclonal antibody reacts with human CD25, which is also known as the low-affinity interleukin (IL)-2 receptor alpha. CD25 is expressed on activated T cells (including a subset of regulatory T cells), B cells, and macrophages. This receptor subunit associates with CD122, the IL-2 receptor beta chain, and CD132, the common gamma chain, to form the high-affinity IL-2 receptor. IL-2 receptor signaling has been linked to cell proliferation and survival in lymphocytes. CD25 is a marker for Hodgkin's disease, and has been associated with such autoimmune diseases such as Type I diabetes.Applications Reported: This GLLZDMY antibody has been reported for use in western blotting, and immunohistochemical staining of formalin-fixed paraffin embedded tissue sections.Applications Tested: This GLLZDMY antibody has been tested by immunohistochemistry of formalin-fixed paraffin embedded tissue using either low or high pH antigen retreival and can be used at 5 μg/mL. It is recommended that the antibody be carefully titrated for opitmal performance in the assay of interest.Purity: Greater than 90%, as determined by SDS-PAGE.Aggregation: Less than 10%, as determined by HPLC.Filtration: 0.2μm post-manufacturing filtered.
Specifications

Specifications

CD25
Monoclonal
0.5mg/mL
PBS with 0.09% sodium azide; pH 7.2
P01589
IL2 RA antibody, Il2ra antibody, Interleukin 2 receptor alpha chain antibody, TAC antigen antibody, IL-2R alpha chain
Murine
Affinity chromatography
RUO
Antibody
Monoclonal
Human
Immunohistochemistry (Paraffin), Western Blot
GLLZDMY
Unconjugated
IL2RA
Liquid
IL2RA
IgG1, kappa, kappa
100μg
Store at 2-8°C.
Primary
3559
Videos
Documents

For Research Use Only.