missing translation for 'onlineSavingsMsg'
Learn More
Learn More
CD25/IL-2Rα Ab-1 Mouse Monoclonal Antibody, Epredia™
Obtain accurate, reproducible results in immunofluorescence and flow cytometry experiments with Epredia™ CD25/IL-2Rα Ab-1, Mouse Monoclonal Antibody.
£113.00 - £447.78
Specifications
Antigen | CD25/IL-2Ralpha Ab-1 |
---|---|
Clone | IL2R.1 |
Applications | Flow Cytometry, Immunofluorescence |
Classification | Monoclonal |
Conjugate | Unconjugated |
Product Code | Brand | Concentration | Quantity | Price | Quantity & Availability | ||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Concentration | Quantity | Price | Quantity & Availability | ||||
12680527
|
Epredia
MS-203-P0 |
200μg/mL | 100 μL |
£113.00
100µL |
Estimated Shipment: 03-05-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | |||
12690527
|
Epredia
MS-203-P1 |
200μg/mL | 500 μL |
£313.00
500µL |
Estimated Shipment: 03-05-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | |||
12600537
|
Epredia
MS-203-P1ABX |
1mg/mL | 100 μL |
£357.00
100µL |
Estimated Shipment: 03-05-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | |||
12670527
|
Epredia
MS-203-P |
200μg/mL | 1 mL |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
|||||
12610537
|
Epredia
MS-203-PABX |
1mg/mL | 200 μL |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
|||||
Description
IL-2R (T-cell growth factor receptor) consists of a chain of 55kDa (also known as Tac antigen or p55 or α chain) and a p75 or Β chain. IL-2Rα is over-expressed on the surface of activated T-cells, HTLV-1 or -2 positive T-cell lines, and moderately expressed on activated B-cells, NK-cells, and macrophages.Host Species: Mouse
Clone: IL2R.1
Isotype: IgG1/κ
Species Reactivity: Human. Others not tested.
Epitope: Not determined
Immunogen: Recombinant human interleukin-2 receptor
Molecular Weight: 55 kDa
Positive Control: Tonsil or lymph node
Cellular Localization: Membrane
Recommended for:
- Immunofluorescence
- Flow Cytometry
Note:
USA: RUO; Int'l: RUO
Specifications
CD25/IL-2Ralpha Ab-1 | |
Flow Cytometry, Immunofluorescence | |
Unconjugated | |
Mouse | |
B-Cell | |
Recombinant human interleukin-2 receptor | |
Primary |
IL2R.1 | |
Monoclonal | |
Concentrated | |
RUO | |
Human | |
IgG2a κ |