missing translation for 'onlineSavingsMsg'
Learn More

CD244 Mouse anti-Human, FITC, Clone: eBioDM244, eBioscience™

Mouse Monoclonal Antibody

Brand:  Affymetrix eBioscience 11-5837-41

Code : NEW

Additional Details : Weight : 0.23750kg

 View more versions of this product

Product Code. 15596566

  • £65.75 / 25 tests

Estimated Shipment
23-10-2020

Please to purchase this item. Need a web account? Register with us today!


Description

Description

CD244 (2B4, SLAMF4) is a cell surface receptor expressed on natural killer (NK) cells (and some T cells) that mediate non-major histocompatibility complex (MHC) restricted killing. The interaction between NK-cell and target cells via CD244 is thought to modulate NK-cell cytolytic activity. CD244 interacts with SLAMF2, causing the activation of both SLAMF4- and SLAMF2-expressing cells. Patients with systemic lupus erythmatosus have lower than normal levels of SLAMF4 expressed on their NK cells and monocytes, suggesting that SLAMF4 may play a role in the pathology of this autoimmune disease. Diseases associated with CD244 dysfunction include rheumatoid arthritis and lymphoproliferative syndrome. Alternatively spliced transcript variants encoding different isoforms of CD244 have been found for this gene.
Specifications

Specifications

CD244
Monoclonal
5 μL/Test
PBS with 0.1% gelatin, 0.2% BSA and 0.09% sodium azide; pH 7.2
Q9BZW8
2B4, NAIL, NKR2B4, Nmrk, SLAMF4
Mouse
Affinity chromatography
RUO
Antibody
Monoclonal
Human
Flow Cytometry
eBioDM244
FITC
CD244
Liquid
CD244
IgG1, kappa
25 Tests
4° C, store in dark, DO NOT FREEZE!
Primary
51744
Videos
Documents

For Research Use Only.