missing translation for 'onlineSavingsMsg'
Learn More

CD244, clone: eBioPP35 (PP35), eBioscience™

Mouse Monoclonal Antibody

Brand:  Affymetrix eBioscience 14-2449-82

Code : Z2

Additional Details : Weight : 0.01000kg

 View more versions of this product

Product Code. 15237327

  • £68.21 / 100µg


Call for latest availability
+44 (0)1509 555 500
Add to basket

Description

Description

Description: The eBioPP35 monoclonal antibody reacts with human CD244, also known as 2B4, p38, NK-cell activation-inducing ligand, signaling lymphocyte activation molecule family member 4, and natural killer cell receptor 2B4. Human CD244 is a 38 kD protein expressed on NK cells, a subset of CD8+ T cells, gammadelta T cells, monocytes, basophils, mast cells, and eosinophils. Binding of CD244 to its ligand, CD48, results in NK cell activation. Costaining experiments with eBioC1.7 suggest that both antibodies recognize different epitopes.Applications Reported: This eBioPP35 (PP35) antibody has been reported for use in flow cytometric analysis, immunoprecipitation, and immunoblotting (WB).Purity: Greater than 90%, as determined by SDS-PAGE.Aggregation: Less than 10%, as determined by HPLC.Filtration: 0.2μm post-manufacturing filtered.
Specifications

Specifications

CD244
Monoclonal
0.5mg/mL
PBS with 0.09% sodium azide; pH 7.2
Q9BZW8
2B4, NAIL, NKR2B4, Nmrk, SLAMF4
Murine
Affinity chromatography
RUO
Antibody
Monoclonal
Human
Flow CytometryþImmunoprecipitationþWestern Blot
eBioPP35 (PP35)
Unconjugated
CD244
Purified
CD244
IgG1
100μg
Store at 2-8°C.
Primary
51744
Videos

For Research Use Only.