CD244 Mouse anti-Human, APC, Clone: eBioDM244, eBioscience™

Mouse Monoclonal Antibody

Overview
Brand: Affymetrix eBioscience

Manufacturer Part Number: 17-5837-41

25TEST Anti-Human CD244 APC

Code: NEW

Additional Details:
Additional Details: Weight: 0.23750kg



Disclaimers: For Research Use Only.

Product Code. 15528146

Quantity Price
1 £ 84.99 / 25 tests
Estimated Shipment date
from Supplier 27-02-2017
Add to basket


Description and Specification

Specification

Antigen CD244
Format Conjugated
Formulation aqueous buffer, 0.09% sodium azide, may contain carrier protein/stabilizer
Gene Alias 2B4, p38, SLAMF4
Host Species Mouse
Applications Flow Cytometry
Isotype IgG1
Quantity 25 tests
Regulatory Status RUO
Species Reactivity Human
Storage Requirements Store at 2-8°C. Do not freeze. Light-sensitive material.
Primary or Secondary Primary
Monoclonal or Polyclonal Monoclonal
Clone eBioDM244
Concentration 5μL (0.125μg)/test
Conjugate APC

The eBioDM244 monoclonal antibody reacts with human CD244 (2B4, p38). In human, CD244 is a 38kDa protein expressed on NK cells, a subset of CD8+ T cells, γδ T cells, monocytes, basophils and eosinophils. Binding of the CD244 ligand, CD48, results in NK cell activation, unlike mouse CD244, which is an inhibitory receptor.