missing translation for 'onlineSavingsMsg'
Learn More

CD244.2 (2B4), clone: eBio244F4, eBioscience™

Rat Monoclonal Antibody

Brand:  Affymetrix eBioscience 14-2441-81

Code : Z2

Additional Details : Weight : 0.01000kg

 View more versions of this product

Product Code. 15297317

  • £44.53 / 50µg


Estimated Shipment
01-04-2019
Add to basket

Description

Description

Applications Tested: This eBio244F4 antibody has been tested by flow cytometric analysis of IL-2-stimulated mouse splenocytes. This can be used at less than or equal to 0.06 μg per test. A test is defined as the amount (μg) of antibody that will stain a cell sample in a final volume of 100 μL. Cell number should be determined empirically but can range from 10^5 to 10^8 cells/test. It is recommended that the antibody be carefully titrated for optimal performance in the assay of interest. Purity: Greater than 90%, as determined by SDS-PAGE. Aggregation: Less than 10%, as determined by HPLC. Filtration: 0.2 μm post-manufacturing filtered. This gene encodes a cell surface receptor expressed on natural killer (NK) cells (and some T cells) that mediate non-major histocompatibility complex (MHC) restricted killing. The interaction between NK-cell and target cells via this receptor is thought to modulate NK-cell cytolytic activity. Alternatively spliced transcript variants encoding different isoforms have been found for this gene.
Specifications

Specifications

CD244.2 (2B4)
Monoclonal
0.5 mg/mL
PBS with 0.09% sodium azide; pH 7.2
Q07763
B6 Alloantigen
Rat
Affinity chromatography
RUO
Antibody
Monoclonal
Mouse
Flow Cytometry, Western Blot
eBio244F4
Unconjugated
Cd244
Liquid
Cd244
IgG2a
50 μg
4° C
Primary
18106
Videos

For Research Use Only.