missing translation for 'onlineSavingsMsg'
Learn More

CD244.1, PerCP-eFluor™ 710, clone: C9.1, eBioscience™

Rat Monoclonal Antibody

Brand:  Affymetrix eBioscience 46-2440-80

Code : NEW

Additional Details : Weight : 0.09500kg

 View more versions of this product

Product Code. 15529226

  • £93.27 / 25µg


Estimated Shipment date
from Supplier 28-06-2018
Add to basket

Description

Description

Description: The monoclonal antibody C9.1 reacts with mouse CD244.1. This receptor is expressed on NK cells, gamma delta T cells, some CD8+ T cells, basophils, and monocytes on most inbred mouse strains except C57BL/6 and C58, which express the CD244.2 allotype. CD244 is a CD2-related receptor belonging to the SLAM family that exists as two different isoforms with opposing functions. CD244 associates with CD48, which is expressed widely on hematopoietic cells, regulates NK cell proliferation, cytotoxicity, and cytokine production. However, the CD244/CD28 interaction between NK and other cell types inhibits NK cytotoxicity, which contributes to self-tolerance. CD244 may enhance or inhibit CD8+ T cell effector function depending on SAP levels.The C9.1 antibody has been reported to have antagonist activity on activated NK cells.Applications Reported: This C9.1 antibody has been reported for use in flow cytometric analysis.Applications Tested: This C9.1 antibody has been tested by flow cytometric anaylsis of BALB/c mouse splenocytes. This can be used at less than or equal to 0.125 μg per test. A test is defined as the amount (μg) of antibody that will stain a cell sample in a final volume of 100 μL. Cell number should be determined empirically but can range from 10^5 to 10^8 cells/test. It is recommended that the antibody be carefully titrated for optimal performance in the assay of interest.PerCP-eFluor® 710 emits at 710 nm and is excited with the blue laser (488 nm); it can be used in place of PerCP-Cyanine5.5. We recommend using a 710/50 bandpass filter, however, the 695/40 bandpass filter is an acceptable alternative. Please make sure that your instrument is capable of detecting this fluorochrome.Fixation: Samples can be stored in IC Fixation Buffer (cat. 00-8222) (100 μL of cell sample + 100 μL of IC Fixation Buffer) or 1-step Fix/Lyse Solution (cat. 00-5333) for up to 3 days in the dark at 4°C with minimal impact on brightness and FRET efficiency/compensation. Some generalizations regarding fluorophore performance after fixation can be made, but clone specific performance should be determined empirically.Excitation: 488 nm; Emission: 710 nm; Laser: Blue Laser.Filtration: 0.2μm post-manufacturing filtered.
Specifications

Specifications

CD244.1
Monoclonal
0.2mg/mL
PBS with 0.1% gelatin and 0.09% sodium azide; pH 7.2
Q07763
2B4, SLAMF4, NK cell type I receptor protein
Rat
Affinity chromatography
RUO
Antibody
Monoclonal
Murine
Flow Cytometry
C9.1
PerCP-eFluor 710
Cd244
Conjugated
Cd244
IgG2b, kappa
25μg
Store at 2-8°C. Do not freeze. Light-sensitive material.
Primary
18106
Videos

For Research Use Only.