missing translation for 'onlineSavingsMsg'
Learn More

CD244.1 Monoclonal Antibody (C9.1), APC, eBioscience™, Invitrogen™

Rat Monoclonal Antibody

Brand:  Affymetrix eBioscience 17-2440-82

Code : NEW

Additional Details : Weight : 0.09500kg

 View more versions of this product

Product Code. 15548066

  • £247.00 / 100µg
Estimated Shipment: 03-06-2024
to see stock.

Please to purchase this item. Need a web account? Register with us today!


Description

Description

Description: The monoclonal antibody C9.1 reacts with mouse CD244.1. This receptor is expressed on NK cells, gamma delta T cells, some CD8+ T cells, basophils, and monocytes on most inbred mouse strains except C57BL/6 and C58, which express the CD244.2 allotype. CD244 is a CD2-related receptor belonging to the SLAM family that exists as two different isoforms with opposing functions. CD244 associates with CD48, which is expressed widely on hematopoietic cells, regulates NK cell proliferation, cytotoxicity, and cytokine production. However, the CD244/CD28 interaction between NK and other cell types inhibits NK cytotoxicity, which contributes to self-tolerance. CD244 may enhance or inhibit CD8+ T cell effector function depending on SAP levels. The C9.1 antibody has been reported to have antagonist activity on activated NK cells. Applications Reported: This C9.1 antibody has been reported for use in flow cytometric analysis. Applications Tested: This C9.1 antibody has been tested by flow cytometric analysis of mouse splenocytes. This can be used at less than or equal to 0.25 μg per test. A test is defined as the amount (μg) of antibody that will stain a cell sample in a final volume of 100 μL. Cell number should be determined empirically but can range from 10^5 to 10^8 cells/test. It is recommended that the antibody be carefully titrated for optimal performance in the assay of interest. Excitation: 633-647 nm; Emission: 660 nm; Laser: Red Laser.

CD244.1 Monoclonal antibody specifically detects CD244.1 in Mouse samples. It is validated for Flow Cytometry.
Specifications

Specifications

CD244.1
Monoclonal
0.2 mg/mL
PBS with 0.09% sodium azide; pH 7.2
Q07763
Cd244
Affinity chromatography
RUO
18106
4° C, store in dark, DO NOT FREEZE!
Liquid
Flow Cytometry
C9.1
APC
Cd244
2B4; C9.1; CD244; CD244 long isoform; CD244 natural killer cell receptor 2B4; CD244 short isoform; F730046O15Rik; Ly90; NAIL; natural killer cell receptor; Natural killer cell receptor 2B4; NK cell receptor 2B4; NK cell type I receptor protein 2B4; NKR2B4; Nmrk; non MHC restricted killing associated; non-MHC restricted killing associated; Signaling lymphocytic activation molecule 4; SLAM family member 4; SLAMF4
Rat
100 μg
Primary
Mouse
Antibody
IgG2b κ
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers

For Research Use Only.