missing translation for 'onlineSavingsMsg'
Learn More

CD24, PE, clone: 30-F1, eBioscience™

Rat Monoclonal Antibody

Brand:  Affymetrix eBioscience 12-0241-81

Code : Z2

Additional Details : Weight : 0.25000kg

 View more versions of this product

Product Code. 15238489

  • £49.52 / 50µg


Estimated Shipment date
from Supplier 03-07-2018
Add to basket

Description

Description

Description: The 30-F1 monoclonal antibody reacts with the mouse CD24 molecule, also known as Heat Stable Antigen (HSA). This 35-50kDa molecule is anchored in the plasma membrane via phosphatidylinositol and is expressed by erythrocytes, thymocytes, peripheral lymphocytes and myeloid lineage. The expression of CD24 detected by 30-F1 has been used to resolve stages of B lymphopoiesis in mouse bone marrow. It has been reported that P-selectin (CD62P) binds to CD24. CD24 is a variably glycosylated molecule resulting in heterogeneity of molecular mass of this antigen on cells of different lineages and different antibodies to CD24 exhibit subtle differences in staining level on lymphocyte populations.Applications Reported: The 30-F1 antibody has been reported for use in flow cytometric analysis.Applications Tested: The 30-F1 antibody has been tested by flow cytometric analysis of mouse splenocytes and bone marrow cells. This can be used at less than or equal to 0.125 μg per test. A test is defined as the amount (μg) of antibody that will stain a cell sample in a final volume of 100 μL. Cell number should be determined empirically but can range from 10^5 to 10^8 cells/test. It is recommended that the antibody be carefully titrated for optimal performance in the assay of interest.Excitation: 488-561 nm; Emission: 578 nm; Laser: Blue Laser, Green Laser, Yellow-Green Laser.Filtration: 0.2μm post-manufacturing filtered.
Specifications

Specifications

CD24
Monoclonal
0.2mg/mL
PBS with 0.1% gelatin and 0.09% sodium azide; pH 7.2
P24807
CD24 antigen (small cell lung carcinoma cluster 4 antigen), CD24A, FLJ22950, FLJ43543, MGC75043
Rat
Affinity chromatography
RUO
Antibody
Monoclonal
Murine
Flow Cytometry
30-F1
PE
Cd24a
Conjugated
Cd24a
IgG2c, kappa
50μg
Store at 2-8°C. Do not freeze. Light-sensitive material.
Primary
12484
Videos

For Research Use Only.