missing translation for 'onlineSavingsMsg'
Learn More

CD24, clone: M1/69, eBioscience™

Rat Monoclonal Antibody

Brand:  Affymetrix eBioscience 14-0242-81

Code : Z2

Additional Details : Weight : 0.01000kg

 View more versions of this product

Product Code. 15206867

  • £36.94 / 50µg


Call for latest availability
+44 (0)1509 555 500
Add to basket

Description

Description

Description: The M1/69 monoclonal antibody reacts with the mouse CD24 molecule, also known as Heat Stable Antigen (HSA). This 35-50kDa molecule is anchored in the plasma membrane via phosphatidylinositol and is expressed by erythrocytes, thymocytes, peripheral lymphocytes and myeloid lineage. CD24 is a variably glycosylated molecule resulting in heterogeneity of molecular mass of this antigen on cells of different lineages and antibodies to CD24 exhibit subtle differences in staining level on lymphocyte populations. The expression of CD24 has been used to resolve stages of B lymphopoiesis in mouse bone marrow. It has been reported that P-selectin (CD62P) binds to CD24.Applications Reported: The M1/69 antibody has been reported for use in flow cytometric analysis, immunoblotting (WB), and immunohistochemical staining. M1/69 has also been reported in in vitro functional assays. (Please use Functional Grade purified M1/69, cat. 16-0242, in functional assays.).Applications Tested: The M1/69 antibody has been tested by flow cytometric analysis of mouse splenocytes and thymocytes. This can be used at less than or equal to 0.125 μg per test. A test is defined as the amount (μg) of antibody that will stain a cell sample in a final volume of 100 μL. Cell number should be determined empirically but can range from 10^5 to 10^8 cells/test. It is recommended that the antibody be carefully titrated for optimal performance in the assay of interest.Purity: Greater than 90%, as determined by SDS-PAGE.Aggregation: Less than 10%, as determined by HPLC.Filtration: 0.2μm post-manufacturing filtered.
Specifications

Specifications

CD24
Monoclonal
0.5mg/mL
PBS with 0.09% sodium azide; pH 7.2
P24807
CD24 antigen (small cell lung carcinoma cluster 4 antigen), CD24A, FLJ22950, FLJ43543, MGC75043
Rat
Affinity chromatography
RUO
Antibody
Monoclonal
Murine
Western Blot
M1/69
Unconjugated
Cd24a
Purified
Cd24a
IgG2b, kappa
50μg
Store at 2-8°C.
Primary
12484
Videos

For Research Use Only.