missing translation for 'onlineSavingsMsg'
Learn More

CD24, APC-eFluor 780, clone: M1/69, eBioscience™

Rat Monoclonal Antibody

Brand:  Affymetrix eBioscience 47-0242-82

Code : Z2

Additional Details : Weight : 0.25000kg

 View more versions of this product

Product Code. 15381610

  • £159.14 / 100µg


EU Stock 2
Add to basket

Description

Description

Description: The M1/69 monoclonal antibody reacts with the mouse CD24 molecule, also known as Heat Stable Antigen (HSA). This 35-50kDa molecule is anchored in the plasma membrane via phosphatidylinositol and is expressed by erythrocytes, thymocytes, peripheral lymphocytes and myeloid lineage. CD24 is a variably glycosylated molecule resulting in heterogeneity of molecular mass of this antigen on cells of different lineages and antibodies to CD24 exhibit subtle differences in staining level on lymphocyte populations. The expression of CD24 has been used to resolve stages of B lymphopoiesis in mouse bone marrow. It has been reported that P-selectin (CD62P) binds to CD24.Applications Reported: This M1/69 antibody has been reported for use in flow cytometric analysis.Applications Tested: This M1/69 antibody has been tested by flow cytometric analysis of mouse splenocytes. This can be used at less than or equal to 0.125 μg per test. A test is defined as the amount (μg) of antibody that will stain a cell sample in a final volume of 100 μL. Cell number should be determined empirically but can range from 10^5 to 10^8 cells/test. It is recommended that the antibody be carefully titrated for optimal performance in the assay of interest.APC-eFluor® emits at 780 nm and is excited with the Red laser (633 nm). Please make sure that your instrument is capable of detecting this fluorochome.Light sensitivity: This tandem is sensitive to photo-induced oxidation. Please protect this vial and stained samples from light. Fixation: Samples can be stored in IC Fixation Buffer (cat. 00-8222) (100 μL cell sample + 100 μL IC Fixation Buffer) or 1-step Fix/Lyse Solution (cat. 00-5333) for up to 3 days in the dark at 4°C with minimal impact on brightness and FRET efficiency/compensation. Some generalizations regarding fluorophore performance after fixation can be made, but clone specific performance should be determined empirically.Excitation: 633-647 nm; Emission: 780 nm; Laser: Red Laser.Filtration: 0.2μm post-manufacturing filtered.
Specifications

Specifications

CD24
Monoclonal
0.2mg/mL
PBS with 0.09% sodium azide; pH 7.2
P24807
CD24 antigen (small cell lung carcinoma cluster 4 antigen), CD24A, FLJ22950, FLJ43543, MGC75043
Rat
Affinity chromatography
RUO
Antibody
Monoclonal
Murine
Flow Cytometry
M1/69
APC-eFluor 780
Cd24a
Conjugated
Cd24a
IgG2b, kappa
100μg
Store at 2-8°C. Do not freeze. Light-sensitive material. This tandem dye is sensitive to photo-induced oxidation. Protect this vial from light during storage, handling and experimental procedures.
Primary
12484
Videos

For Research Use Only.