missing translation for 'onlineSavingsMsg'
Learn More

CD24 Mouse anti-Human, eFluor® 450, Clone: eBioSN3 (SN3 A5-2H10), eBioscience™

Mouse Monoclonal Antibody

Brand:  Affymetrix eBioscience 48-0247-41

Code : NEW

Additional Details : Weight : 0.23750kg

Product Code. 15579646

  • £79.25 / 25 tests

Estimated Shipment
03-11-2020

Please to purchase this item. Need a web account? Register with us today!


Description

Description

CD24 is a two-chain glycosylphosphatidylinositol (GPI)-anchored glycoprotein expressed at multiple stages of B-cell development, beginning with the bone marrow pro-B-cell compartment and continuing through mature, surface Ig positive B-cells. Plasma cell expression is very low or negative. CD24 is also expressed on the majority of B-lineage acute lymphoblastic leukemias, B-cell CCLs and B-cell non-Hodgkin′s lymphomas. CD24 may play a role in regulation of B-cell proliferation and maturation, and control of autoimmunity.
Specifications

Specifications

CD24
Monoclonal
5 μL/Test
PBS with 0.1% gelatin, 0.2% BSA and 0.09% sodium azide; pH 7.2
P25063
CD24 antigen (small cell lung carcinoma cluster 4 antigen); CD24A; FLJ22950; FLJ43543; MGC75043
Mouse
Affinity chromatography
RUO
Antibody
Monoclonal
Human
Flow Cytometry
eBioSN3 (SN3 A5-2H10)
eFluor™ 450
CD24
Liquid
CD24
IgG1, kappa
25 Tests
4° C, store in dark, DO NOT FREEZE!
Primary
100133941
Videos
Documents

For Research Use Only.